Skip to Content
Merck

Skip To

L7269

α-Lactalbumin from human milk

≥95% (SDS-PAGE), lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.61
MDL number:
Biological source:
human milk
Assay:
≥95% (SDS-PAGE)
Form:
lyophilized powder
Technique(s):
cell migration: suitable
Concentration:
>75 % protein (UV)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human milk

Quality Level

assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

14,070 Da by calculation

concentration

>75 % protein (UV)

technique(s)

cell migration: suitable

solubility

H2O: soluble 10 mg/mL(lit.)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... LALBA(3906)

General description

α-Lactalbumin (α-LA) is a small, acidic,[1] whey protein that constitutes about 22% of the total proteins in human milk. It is produced by the epithelial cells of the mammary gland. α-LA is made up of two domains, a large α-helical domain, and a small β-sheet domain.[1]

Application

α-Lactalbumin (α-LA) has been used as a standard

  • to study the partitioning behavior of different monomeric proteins with exposure to amino acids on the protein surface[2]
  • to study the interaction between α-LA and cathepsin D
  • to study the ability of breast milk fractions to enhance the transepithelial flux of extrinsic iron in colon carcinoma cell line

Biochem/physiol Actions

α-Lactalbumin (α-LA) forms a complex with lactose synthase within the mammary gland and plays a role in milk production and regulates milk volume. It acts as an essential source for bioactive peptides and essential amino acids such as lysine, tryptophan, branched-chain amino acids, and sulfur-containing amino acids that play a role in an infant′s nutrition. In addition, α-LA has a wide range of applications including a supplement to foster gastrointestinal health and modulate sleep and depression. α-LA also shows therapeutic effects against sarcopenia, seizures, mood disorders, and cancer. It has a Ca2+ binding site that binds with Na+, K+, Mg2+, and Mn2+ and many Zn2+ binding sites.[1]
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase.
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase. Complexes of α-lactalbumin with oleic acid show drastically different activities than α-lactalbumin alone, being strongly cytotoxic to tumor cells. The complex is referred to as HAMLET (human alpha-lactalbumin made lethal to tumor cells).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L6385L72521013909
technique(s)

cell migration: suitable

technique(s)

electrophoresis: suitable

technique(s)

cell culture | insect: suitable

technique(s)

-

biological source

human milk

biological source

bovine milk

biological source

bovine

biological source

-

assay

≥95% (SDS-PAGE)

assay

-

assay

-

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-

form

lyophilized powder

form

powder

form

powder

form

-

Gene Information

human ... LALBA(3906)

Gene Information

cow ... LALBA(281894)

Gene Information

bovine ... LALBA(281894)

Gene Information

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Breast Milk Fractions Solubilize Fe(III) and Enhance Iron Flux across Caco-2 Cells
Robert E. S.
The Journal of Nutrition, 449?455-449?455 (2003)
HAMLET effect on cell death and mitochondrial respiration in colorectal cancer cell lines with KRAS/BRAF mutations.
??ilinskas, et al.
Journal of Cancer Research and Clinical Oncology (2023)
Applications for ?-lactalbumin in human nutrition
Donald K L, et al.
Nutrition Research Reviews, 76(6), 444?460-444?460 (2018)



Global Trade Item Number

SKUGTIN
L7269-25MG04061833965436
L7269-5MG04061833965443

Questions

Reviews

No rating value

Active Filters