Skip to Content
Merck

Skip To

I6383

Insulin Chain B Oxidized from bovine pancreas

≥80% (HPLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C157H232N40O47S2
CAS Number:
Molecular Weight:
3495.89
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Biological source:
bovine
Form:
powder
Assay:
≥80% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Level

sterility

non-sterile

assay

≥80% (HPLC)

form

powder

mol wt

3496 Da by calculation (Average Mass)

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

[S](=O)(=O)(O)C[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)Cc8c

InChI

1S/C157H232N40O47S2/c1-79(2)57-104(181-130(211)86(15)172-136(217)103(50-53-125(209)210)180-152(233)127(84(11)12)194-148(229)107(60-82(7)8)184-145(226)113(67-95-70-165-78-171-95)189-150(231)115(74-198)176-123(206)73-169-133(214)116(75-245(239,240)241)191-140(221)105(58-80(3)4)182-144(225)112(66-94-69-164-77-170-94)188-138(219)102(48-51-119(160)202)178-146(227)114(68-120(161)203)190-153(234)126(83(9)10)193-131(212)98(159)61-89-31-21-18-22-32-89)139(220)185-110(64-92-40-44-96(200)45-41-92)142(223)183-106(59-81(5)6)147(228)195-128(85(13)14)154(235)192-117(76-246(242,243)244)134(215)168-71-121(204)174-101(49-52-124(207)208)137(218)177-99(38-29-55-166-157(162)163)132(213)167-72-122(205)175-108(62-90-33-23-19-24-34-90)141(222)186-109(63-91-35-25-20-26-36-91)143(224)187-111(65-93-42-46-97(201)47-43-93)149(230)196-129(88(17)199)155(236)197-56-30-39-118(197)151(232)179-100(37-27-28-54-158)135(216)173-87(16)156(237)238/h18-26,31-36,40-47,69-70,77-88,98-118,126-129,198-201H,27-30,37-39,48-68,71-76,158-159H2,1-17H3,(H2,160,202)(H2,161,203)(H,164,170)(H,165,171)(H,167,213)(H,168,215)(H,169,214)(H,172,217)(H,173,216)(H,174,204)(H,175,205)(H,176,206)(H,177,218)(H,178,227)(H,179,232)(H,180,233)(H,181,211)(H,182,225)(H,183,223)(H,184,226)(H,185,220)(H,186,222)(H,187,224)(H,188,219)(H,189,231)(H,190,234)(H,191,221)(H,192,235)(H,193,212)(H,194,229)(H,195,228)(H,196,230)(H,207,208)(H,209,210)(H,237,238)(H4,162,163,166)(H,239,240,241)(H,242,243,244)/t86-,87-,88+,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,126-,127-,128-,129-/m0/s1

InChI key

CZJQESISDHGFTC-DQCCOYHDSA-N

Gene Information

cow ... INS(280829)

General description

Insulin is made up of two polypeptide chains (A and B) linked together by two disulfide linkages. Chain A and B are composed of 21 amino acids and 30 amino acids, respectively.

Application

Insulin Chain B Oxidized from bovine pancreas has been used:
  • as a matrix-assisted laser desorption/ionization (MALDI)-mass spectrometry (MS) calibration standard[1][2]
  • in electrochemical impedance spectroscopy and quartz crystal nanobalance (EQCN) studies of insulin adsorption on platinum
  • as an internal standard to quantify insulin chain B residue in protein samples[3]

Biochem/physiol Actions

Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids. It inhibits the breakdown of glycogen, protein, and fat. The β chain is a substrate for carboxypeptidase Y.

Preparation Note

Prepared by modification of Sanger, F., et al.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I1764I1633I5500
biological source

bovine

biological source

-

biological source

bovine

biological source

-

assay

≥80% (HPLC)

assay

-

assay

≥80% (HPLC)

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

-

form

powder

form

powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

Gene Information

cow ... INS(280829)

Gene Information

cow ... INS(280829)

Gene Information

cow ... INS(280829)

Gene Information

cow ... INS(280829)


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Electrochemical impedance spectroscopy and quartz crystal nanobalance (EQCN) studies of insulin adsorption on Pt
Wright JEI, et al.
Journal of Electroanalytical Chemistry, 564, 185-197 (2004)
Johansen, J.T., et al.
Carlsberg Research Communications, 41, 1-1 (1976)
Epitope mapping by a combination of epitope excision and MALDI-MS.
C E Parker et al.
Methods in molecular biology (Clifton, N.J.), 146, 185-201 (2000-08-19)



Global Trade Item Number

SKUGTIN
I6383-10MG04061832820712
I6383-5MG04061832820736
I6383-1MG04061826284865
I6383-50MG04061832820729

Questions

Reviews

No rating value

Active Filters