Skip to Content
Merck

Skip To

I6379

(R)-(−)-Isoproterenol

crystalline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C11H17NO3
CAS Number:
Molecular Weight:
211.26
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Form:
crystalline
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

crystalline

Quality Segment

color

white to yellow

storage temp.

−20°C

SMILES string

CC(C)NC[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C11H17NO3/c1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8/h3-5,7,11-15H,6H2,1-2H3/t11-/m0/s1

InChI key

JWZZKOKVBUJMES-NSHDSACASA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7257N7251C8128
form

crystalline

form

crystalline

form

crystalline

form

crystalline

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

color

white to yellow

color

off-white to tan

color

yellow

color

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
I6379-1G04061825982373
I6379-500MG04061832072067
I6379-5G04061832091532
I6379-100MG04061832072050

Questions

  1. What can be used to make a solution of (R)-(−)-Isoproterenol, Product I6379?

    1 answer
    1. As mentioned on the product specification sheet, this chemical is soluble in 1 N HCl (50 mg/ml). Please note that this product may contain insoluble matter upon solubilizing. Please view the product specification sheet for the requested information: https://www.sigmaaldrich.com/specification-sheets/963/173/I6379-BULK________SIGMA____.pdf

      Helpful?

Reviews

No rating value

Active Filters