Skip to Content
Merck
All Photos(1)

Key Documents

H0146

Sigma-Aldrich

Horse serum

liquid

Synonym(s):

Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
biological source:
horse serum
sterility:
non-sterile
form:
liquid
shipped in:
dry ice
Pricing and availability is not currently available.

biological source

horse serum

Quality Level

sterility

non-sterile

form

liquid

contains

preservative

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Horse serum displays homogeneity between fetal bovine serum and fetal calf serum .It has low levels of polyamine oxidase.[1]
Horse serum is pooled from a normal donor herd.

Application

Horse serum has been used as a supplement in minimum essential medium for cerebrocortical neuronal−glial cell cultures[2] and RPMI-40 medium for PC12 cells derived from rat adrenal medulla pheochromocytoma.[3] It has also been used in immunohistochemistry of Florida manatee brain sections.[4]
Normal horse serum may be used as a blocking agent and controls in immunoassays. For immunohistochemistry and immunocytochemistry, 10% horse serum was used as blocking agent in various Studies.[5][6][7][8][9]
Horse serum was used in immunostaining for blocking non-specific antibodies binding.[10]

Biochem/physiol Actions

The horse serum is an alternative to bovine serum for growth factors and hormones for cell culture studies.[11] Horse serum stimulates fibroblast differentiation and extracellular matrix (ECM) and cytokines synthesis.[11]

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuron types in the presumptive primary somatosensory cortex of the Florida manatee (Trichechus manatus latirostris)
Reyes LD, et al.
Brain, behavior and evolution, 86(3-4), 210-231 (2015)
Coexpression of fractalkine and its receptor in normal human endometrium and in endometrium from users of progestin-only contraception supports a role for fractalkine in leukocyte recruitment and endometrial remodeling
Hannan NJ et al
J. Clin. Endocr., 89, 6119-6129 (2004)
Sevoflurane protects rat mixed cerebrocortical neuronal-glial cell cultures against transient oxygen-glucose deprivation: involvement of glutamate uptake and reactive oxygen species
Canas PT, et al.
Anesthesiology, 105(5), 990-990 (2006)
E Dimitriadis et al.
Reproductive biology and endocrinology : RB&E, 1, 34-34 (2003-05-13)
Embryo implantation, endometrial stromal cell decidualization and formation of a functional placenta are critical processes in the establishment and maintenance of pregnancy. Interleukin (IL)-11 signalling is essential for adequate decidualization in the mouse uterus and IL-11 promotes decidualization in the
Jemma Evans et al.
PloS one, 7(12), e53098-e53098 (2013-01-10)
Synchronous development of the endometrium (to achieve a receptive state) and of the embryo is essential for successful implantation and ongoing pregnancy. Endometrial receptivity exists only for a finite time in a menstrual cycle and the endometrium is refractory to

Protocols

How to stain organoids? A complete step-by-step protocol for immunofluorescent (IF) and immunocytochemical (ICC) staining of organoid cultures using antibodies

How to stain organoids? A complete step-by-step protocol for immunofluorescent (IF) and immunocytochemical (ICC) staining of organoid cultures using antibodies

How to stain organoids? A complete step-by-step protocol for immunofluorescent (IF) and immunocytochemical (ICC) staining of organoid cultures using antibodies

How to stain organoids? A complete step-by-step protocol for immunofluorescent (IF) and immunocytochemical (ICC) staining of organoid cultures using antibodies

Questions

  1. 製薬会社で医薬品の品質試験を行なっている者です。こちらのウマ血清は 日局などの公定書に基づいて製していますでしょうか。可能であれば、製法を開示いただければ幸いです。ご回答よろしくお願いいたします。

    1 answer
    1. The specific manufacturing process is considered proprietary, however this is a pooled serum product from healthy herd animals. Neither the testing, nor the processing, is according to the Japanese Pharmacopoeia or other similar agencies. The product is intended for research use only.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service