Skip to Content
Merck
All Photos(4)

Key Documents

G8772

Sigma-Aldrich

Glass beads, acid-washed

425-600 μm (30-40 U.S. sieve)

Synonym(s):

Acid-Washed Beads, Glass Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102422
NACRES:
NB.22

particle size

425-600 μm (30-40 U.S. sieve)

Related Categories

Application

Acid-washed glass beads have been used:
  • for the extraction of DNA from microalgae
  • in the extraction of fatty acids and proteins from yeast cells
  • to lyse glioma cell lines to determine the dihydrofolate reductase (DHFR) enzyme activity

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daria Jacob et al.
Malaria journal, 16(1), 259-259 (2017-07-01)
Yeast cells represent an established bioreactor to produce recombinant proteins for subunit vaccine development. In addition, delivery of vaccine antigens directly within heat-inactivated yeast cells is attractive due to the adjuvancy provided by the yeast cell. In this study, Pichia
Yeast engineering to express sex pheromone gland genes of the oriental fruit moth, Grapholita molesta
Vatanparast M and Kim Y
Journal of Asia-Pacific Entomology, 22(3), 645-654 (2019)
Agnieszka Anna Rawłuszko-Wieczorek et al.
Epigenetics & chromatin, 11(1), 4-4 (2018-01-27)
Protein posttranslational modifications (PTMs) occur broadly in the human proteome, and their biological outcome is often mediated indirectly by reader proteins that specifically bind to modified proteins and trigger downstream effects. Particularly, many lysine methylation sites among histone and nonhistone
Changyong Cheng et al.
Frontiers in cellular and infection microbiology, 7, 287-287 (2017-07-14)
Microbes employ the thioredoxin system to defend against oxidative stress and ensure correct disulfide bonding to maintain protein function.
A simple and rapid technique for the isolation of DNA from microalgae 1
Fawley M W and Fawley K P
Journal of Phycology, 40(1), 223-225 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service