Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352207
Biological source:
bovine fetus
Origin:
non-USA origin
Sterility:
γ-irradiated, sterile-filtered
Form:
lyophilized powder
Pricing and availability is not currently available.
biological source
bovine fetus
sterility
γ-irradiated, sterile-filtered
form
lyophilized powder
composition
hemoglobin (tested)
origin
non-USA origin
availability
available only in ( Europe, Middle East and Africa only)
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
storage temp.
room temp
General description
Foetal Bovine Serium remains a popular media supplement because it provides a wide array of functions in cell culture. Foetal Bovine Serum delivers nutrients, growth and attachment factors and protects cells from oxidative damage and apoptosis by mechanisms that are difficult to reproduce in serum-free media (SFM) systems.
Application
Endotoxin and hemoglobin tested
Analysis Note
- Endotoxin and hemoglobin tested
- Tested for the presence of bacteria, virus, and mycoplasma
- Triple filtered with 0.1 micron membrane under aseptic conditions
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Seav-Ly Tran et al.
PloS one, 6(9), e22876-e22876 (2011-09-13)
Trypan blue is a dye that has been widely used for selective staining of dead tissues or cells. Here, we show that the pore-forming toxin HlyII of Bacillus cereus allows trypan blue staining of macrophage cells, despite the cells remaining
Ala-Eddine Deghmane et al.
PLoS pathogens, 7(12), e1002403-e1002403 (2011-12-07)
Meningococcal invasive isolates of the ST-11 clonal complex are most frequently associated with disease and rarely found in carriers. Unlike carriage isolates, invasive isolates induce apoptosis in epithelial cells through the TNF-α signaling pathway. While invasive and non-invasive isolates are
Agnieszka Wesołowska et al.
Parasitology research, 110(1), 395-402 (2011-07-19)
Fasciolosis is a hepatic parasitic infection that affects many mammal species and creates a great economic and veterinary problem. Molecular mechanisms of parasite-hepatocyte interactions have not been precisely characterized yet. Therefore, the aim of the study was to investigate alterations