Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available.
General description
The Extract-N-Amp™ PCR Diluent is intended for use with all of the Extract-N-Amp, REDExtract-N-Amp™ and SYBR® Green Extract-N-Amp PCR Kits, except the Extract-N-Amp and REDExtract-N-Amp Blood PCR Kits. It is specifically formulated to dilute extracted DNA samples before PCR. This step may ameliorate the amplification of the marker gene by minimizing chimeric molecule formation and decreasing PCR inhibitor concentrations. Extract-N-Amp™ PCR Diluent is intended to supplement the kit reagents. Use of this reagent prevents usage of kits reagents where many dilutions are required.
Legal Information
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
SYBR is a registered trademark of Thermo Fisher Scientific or its subsidiaries
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| color colorless | color - | color colorless | color colorless |
| shipped in wet ice | shipped in dry ice | shipped in wet ice | shipped in wet ice |
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
DNA template dilution impacts amplicon sequencing-based estimates of soil fungal diversity
<B>Castle SC, et al.</B>
PhytoKeys, 100-107 (2018)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| E8155-500ML | 04061837692192 |
| E8155-100ML | 04061838354228 |





