Skip to Content
Merck

Skip To

E114

EHNA hydrochloride

≥98% (HPLC), powder, adenosine deaminase inhibitor

Synonym(s):

erythro-9-(2-Hydroxy-3-nonyl)­adenine hydrochloride, erythro-9-Amino-β-hexyl-­α-methyl-9H-purine-9-ethanol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H23N5O · HCl
CAS Number:
Molecular Weight:
313.83
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Quality level:
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

EHNA hydrochloride, ≥98% (HPLC)

Quality Level

assay

≥98% (HPLC)

storage condition

desiccated

solubility

DMSO: >10 mg/mL, H2O: >10 mg/mL

SMILES string

Cl[H].CCCCCC[C@H]([C@H](C)O)n1cnc2c(N)ncnc12

InChI

1S/C14H23N5O.ClH/c1-3-4-5-6-7-11(10(2)20)19-9-18-12-13(15)16-8-17-14(12)19;/h8-11,20H,3-7H2,1-2H3,(H2,15,16,17);1H/t10-,11+;/m0./s1

InChI key

VVDXNJRUNJMYOZ-VZXYPILPSA-N

Gene Information

General description

EHNA is an adenosine deaminase inhibitor. It possesses antitumor action.

Application

EHNA hydrochloride has been used to treat microexplant culture to examine the direction and the motor responsible for the movement of autophagosomes in axons. It has also been used in nucleoside supplementation to inhibit the breakdown of deoxyadenosine by the enzyme adenosine deaminase.

Biochem/physiol Actions

Adenosine deaminase inhibitor.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
324630H9414G3796
assay

≥98% (HPLC)

assay

≥97% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

solubility

DMSO: >10 mg/mL, H2O: >10 mg/mL

solubility

water: 25 mg/mL

solubility

DMSO: >10 mg/mL

solubility

H2O: >10 mg/mL

storage condition

desiccated

storage condition

OK to freeze, desiccated (hygroscopic)

storage condition

desiccated

storage condition

desiccated

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140)

Gene Information

-

Gene Information

-

Gene Information

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sarah M Bowers et al.
Pediatric rheumatology online journal, 18(1), 54-54 (2020-07-12)
Human adenosine deaminase 2 (ADA2) is an extracellular enzyme that negatively regulates adenosine-mediated cell signaling by converting adenosine to inosine. Altered ADA2 enzyme activity has been associated with some viral infections and rheumatic diseases. The potential utility of ADA2 as
Enzyme inhibitors. 26. Bridging hydrophobic and hydrophilic regions on adenosine deaminase with some 9-(2-hydroxy-3-alkyl)adenines.
H J Schaeffer et al.
Journal of medicinal chemistry, 17(1), 6-8 (1974-01-01)
Dynein-and activity-dependent retrograde transport of autophagosomes in neuronal axons
Katsumata K, et al.
Autophagy, 6(3), 378-385 (2010)



Global Trade Item Number

SKUGTIN
E114-25MG04061833601402
E114-100MG04061832947488

Questions

Reviews

No rating value

Active Filters