Skip to Content
Merck

Skip To

D9184

Dexamethasone

meets USP testing specifications

Synonym(s):

(11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
UNSPSC Code:
12352212
NACRES:
NA.21
PubChem Substance ID:
EC Number:
200-003-9
Beilstein/REAXYS Number:
2066651
MDL number:
Biological source:
synthetic (organic)
Form:
solid
Assay:
97.0-102.0%
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

agency

USP/NF, meets USP testing specifications

assay

97.0-102.0%

form

solid

mp

262-264 °C (lit.)

shipped in

ambient

storage temp.

2-8°C

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1

InChI key

UREBDLICKHMUKA-CXSFZGCWSA-N

General description

Dexamethasone is an anti-inflammatory glucocorticoid with a range of effects on cell survival, cell signaling and gene expression. It is useful for study of apoptosis, cell signaling pathways and gene expression.

Application

Dexamethasone was used as medium supplement in the following studies:
  • To investigate the osteogenic differentiation and chondrogenic differentiation of bone marrow mesenchymal stem cells (MSCs).[1]
  • In α-MEM (minimum essential medium) for inducing the osteogenic and adipogenic differentiation in mesenchymal stem cells from human bone marrow and umbilical cord blood.[2]
  • For the isolation and characterization of mesenchymal stem cells isolated from 6- to 8-week-old C57BL/6J mice.[3]

Biochem/physiol Actions

Glucocorticoid anti-inflammatory agent
Glucocorticoid anti-inflammatory agent. Regulates T cell survival, growth, and differentiation. Inhibits the induction of nitric oxide synthase.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D175631375D8893
biological source

synthetic (organic)

biological source

-

biological source

-

biological source

synthetic (organic)

assay

97.0-102.0%

assay

≥98% (HPLC)

assay

99.6%

assay

≥80% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

solid

form

powder

form

solid

form

powder

shipped in

ambient

shipped in

-

shipped in

-

shipped in

ambient

Gene Information

human ... ABCB1(5243), CYP3A4(1576), IL4(3565), IL5(3567), NR3C1(2908)
mouse ... Abcb1a(18671), Abcb1b(18669), Ifng(15978), Nos2(18126), Ptgs2(19225), Tnf(21926)
rat ... Ar(24208), Nr3c1(24413), Tnf(24835)

Gene Information

human ... ABCB1(5243), CYP3A4(1576), IL4(3565), IL5(3567), NR3C1(2908)
mouse ... Abcb1a(18671), Abcb1b(18669), Ifng(15978), Nos2(18126), Ptgs2(19225), Tnf(21926)
rat ... Ar(24208), Nr3c1(24413), Tnf(24835)

Gene Information

human ... ABCB1(5243), CYP3A4(1576), IL4(3565), IL5(3567), NR3C1(2908)
mouse ... Abcb1a(18671), Abcb1b(18669), Ifng(15978), Nos2(18126), Ptgs2(19225), Tnf(21926)
rat ... Ar(24208), Nr3c1(24413), Tnf(24835)

Gene Information

human ... ABCB1(5243), CYP3A4(1576), IL4(3565), IL5(3567), NR3C1(2908)
mouse ... Abcb1a(18671), Abcb1b(18669), Ifng(15978), Nos2(18126), Ptgs2(19225), Tnf(21926)
rat ... Ar(24208), Nr3c1(24413), Tnf(24835)


Still not finding the right product?

Explore all of our products under Dexamethasone


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters