Skip to Content
Merck

Skip To

D8900

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham

With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

DMEM Hams F12, DME/F-12, 1:1 mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

sodium pyruvate: 0.055 g/L
HEPES: 15 mM
NaHCO3: no
L-glutamine: yes
glucose: 3.15 g/L
phenol red: yes

shipped in

ambient

storage temp.

2-8°C

Application

Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham is suitable for the growth of a wide range of cell types in low serum conditions. It has been used:

  • as a component of the culture medium for endometrial explants in vitro culture
  • to immerse chitosan-gelatin-glycerol phosphate hydrogels for cytotoxicity evaluation
  • to culture nucleus pulposus (NP) cells to culture human articular chondrocytes for evaluating the toxicity of baicalein

Preparation Note

Formulated to contain 15.6 grams of powder per liter of medium.
Supplement with 1.2 g/L sodium bicarbonate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0547D2906D8437
form

powder

form

powder

form

powder

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

components

sodium pyruvate: 0.055 g/L
HEPES: 15 mM
NaHCO3: no
L-glutamine: yes
glucose: 3.15 g/L
phenol red: yes

components

NaHCO3: no
sodium pyruvate: 0.11 g/L
HEPES: no
phenol red: yes
glucose: 3.15 g/L
L-glutamine: yes

components

NaHCO3: no
L-glutamine: yes
phenol red: no
sodium pyruvate: 0.055 g/L
glucose: 3.15 g/L
HEPES: 15 mM

components

glucose: 3.15 g/L
HEPES: yes
L-glutamine: yes
phenol red: yes
sodium pyruvate: 0.055 g/L

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

400


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Elena Ruggeri et al.
Microscopy and microanalysis : the official journal of Microscopy Society of America, Microbeam Analysis Society, Microscopical Society of Canada, 23(6), 1197-1206 (2017-12-07)
Confocal microscopy was used to image stages of equine zygote development, at timed intervals, after intracytoplasmic sperm injection (ICSI) of oocytes that were matured in vivo or in vitro. After fixation for 4, 6, 8, 12, or 16 h after
Ryosuke Sakumoto et al.
Journal of animal science and biotechnology, 9, 46-46 (2018-06-28)
The aim of the present study was to evaluate CCL8 and CXCL10 expression and its regulatory mechanism in peripheral blood leukocytes (PBLs) at the time of maternal recognition in cows. Blood samples were collected on 14, 15, 16, 17 and
Sang Hwan Kim et al.
PeerJ, 7, e6344-e6344 (2019-02-08)
Here we investigated the expressions of apoptosis-associated genes known to induce programed cell death through mRNA expressions of two matrix metalloproteinases (MMPs) that are involved in the degradation of collagen and basal membrane in luteal cells cultured in the treatment



Global Trade Item Number

SKUGTIN
D8900-10X1L04061833590089
D8900-50L04061835568376
D8900-50K04061832954301
D8900-BIBC04061826573631
D8900-10L04061833590072

Questions

  1. What is the molarity of glucose when reconstituted with water?

    1 answer
    1. Upon reconstitution, the molarity of glucose is 17.5 mM (3.15 g/L).

      Helpful?

Reviews

No rating value

Active Filters