Skip to Content
Merck
All Photos(1)

Documents

D6790

Sigma-Aldrich

Defensin HNP-2 human

≥95% (HPLC)

Synonym(s):

Neutrophil Peptide-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C147H217N43O37S6
CAS Number:
Molecular Weight:
3370.95
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

solid

UniProt accession no.

antibiotic activity spectrum

fungi
mycobacteria
viruses

Mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@@H]2CSSC[C@@H]3NC(=O)[C@H](Cc4ccccc4)NC(=O)[C@H](C)NC(=O)[C@H](Cc5c[nH]c6ccccc56)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](Cc7ccc(O)cc7)NC(=O)[C@@H](NC(=O)[C@H](CSSC[C@H](NC(=O)[C@H](Cc8ccc(O)cc8)NC(=O)[C@@H](N)CSSC[C@H](NC3=O)C(O)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H]([C@@H](C)CC)C(=O)[N@@H]9CCC[C@H]9C(=O)N[C@@H](C)C(=O)N2)NC(=O)[C@@H](NC(=O)CNC(=O)[C@H](Cc%10ccc(O)cc%10)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCC(O)=O)NC(=O)CNC(=O)[C@H](C)NC1=O)[C@@H](C)O)[C@@H](C)CC

InChI

1S/C147H217N43O37S6/c1-13-73(6)114-139(222)168-76(9)118(201)163-63-110(196)170-96(48-50-113(199)200)127(210)172-92(31-22-52-159-145(152)153)125(208)171-93(32-23-53-160-146(154)155)126(209)178-98(58-81-35-41-85(192)42-36-81)123(206)165-65-112(198)186-117(79(12)191)141(224)184-106-70-232-229-67-103-134(217)173-94(33-24-54-161-147(156)157)128(211)189-116(75(8)15-3)142(225)190-55-25-34-108(190)138(221)167-78(11)120(203)181-105(136(219)187-114)69-231-230-68-104(135(218)185-107(143(226)227)71-233-228-66-89(148)121(204)176-100(133(216)182-103)59-82-37-43-86(193)44-38-82)183-132(215)99(57-80-26-17-16-18-27-80)175-119(202)77(10)166-129(212)102(61-84-62-162-90-29-20-19-28-88(84)90)179-130(213)97(56-72(4)5)177-124(207)91(30-21-51-158-144(150)151)169-111(197)64-164-122(205)95(47-49-109(149)195)174-131(214)101(60-83-39-45-87(194)46-40-83)180-140(223)115(74(7)14-2)188-137(106)220/h16-20,26-29,35-46,62,72-79,89,91-108,114-117,162,191-194H,13-15,21-25,30-34,47-61,63-71,148H2,1-12H3,(H2,149,195)(H,163,201)(H,164,205)(H,165,206)(H,166,212)(H,167,221)(H,168,222)(H,169,197)(H,170,196)(H,171,208)(H,172,210)(H,173,217)(H,174,214)(H,175,202)(H,176,204)(H,177,207)(H,178,209)(H,179,213)(H,180,223)(H,181,203)(H,182,216)(H,183,215)(H,184,224)(H,185,218)(H,186,198)(H,187,219)(H,188,220)(H,189,211)(H,199,200)(H,226,227)(H4,150,151,158)(H4,152,153,159)(H4,154,155,160)(H4,156,157,161)/t73-,74-,75-,76-,77-,78-,79+,89-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,114-,115-,116-,117-/m0/s1

InChI key

GRZXCHIIZXMEPJ-HTLKCAKFSA-N

Gene Information

human ... DEFA1(1667)

Amino Acid Sequence

Cys-Tyr-Cys-Arg-Ile-Pro-Ala-Cys-Ile-Ala-Gly-Glu-Arg-Arg-Tyr-Gly-Thr-Cys-Ile-Tyr-Gln-Gly-Arg-Leu-Trp-Ala-Phe-Cys-Cys [Disulfide Bridges: 1−29, 3−18, 8−28]

General description

Chemical structure: peptide

Biochem/physiol Actions

This is an endogenous antibiotic peptide and monocyte chemotactic peptide produced by human neutrophils. Defensins are a family of 3-4 kDa (29-34 amino acids) peptides found in the granules of mammalian phagocytes. The members of this famuly are variably arginine-rich and all share 6 conserved cysteine residues that participate in intramolecular disulfide bonds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S H White et al.
Current opinion in structural biology, 5(4), 521-527 (1995-08-01)
Defensins comprise a structural class of small cationic peptides that exert broad-spectrum antimicrobial activities through membrane permeabilization. Their predominantly beta-sheet structure, stabilized by three disulfide bonds, distinguishes them from other antimicrobial peptides which typically form amphiphilic helices. Defensins bind to
β-Defensins: endogenous antibiotics of the innate host defense response.
Diamond G., and Bevins, C.L.
Clinical Immunology and Immunopathology, 88, 221-225 (1995)
Human β-defensin-2.
Schroder J.M., and Harder, J.
The International Journal of Biochemistry & Cell Biology, 31, 645-651 (1998)
M E Selsted et al.
Infection and immunity, 55(9), 2281-2286 (1987-09-01)
A broad-spectrum antimicrobial peptide present in guinea pig neutrophils was isolated, characterized biochemically, and assessed for microbicidal range and potency in vitro. The guinea pig neutrophil peptide (GPNP) was purified to homogeneity from a granule-rich subcellular fraction of peritoneal exudate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service