Skip to Content
Merck

Skip To

D6134

2-Deoxy-D-glucose

≥99% (GC), crystalline

Synonym(s):

2-Deoxy-D-arabinohexose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
EC Number:
205-823-0
Beilstein/REAXYS Number:
1723331
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Level

assay

≥99% (GC)

form

crystalline

technique(s)

gas chromatography (GC): suitable

color

white

mp

146-147 °C (lit.)

solubility

H2O: soluble 330 mg/mL, clear, colorless

application(s)

advanced drug delivery

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)CC=O

InChI

1S/C6H12O5/c7-2-1-4(9)6(11)5(10)3-8/h2,4-6,8-11H,1,3H2/t4-,5-,6+/m1/s1

InChI key

VRYALKFFQXWPIH-PBXRRBTRSA-N

Application

2-Deoxy-D-glucose (2-DG) is used in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). 2-Deoxy-D-glucose is used in the development of anti-cancer strategies that involve radio- and chemosensitization and oxidative stress.

Biochem/physiol Actions

2-Deoxy-D-Glucose (2-Deoxyglucose) is a glucose analog that inhibits glycolysis via its actions on hexokinase, the rate limiting step of glycolysis. It is phosphorylated by hexokinase to 2-DG-P which can not be further metabolized by phosphoglucose isomerase. This leads to the accumulation of 2-DG-P in the cell and the depletion in cellular ATP. In vitro, 2-Deoxyglucose has been shown to induce autophagy, increase ROS production, and activate AMPK.

Preparation Note

A further purification of the Grade II material by recrystallization.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D8375D3179M9376
assay

≥99% (GC)

assay

≥98% (GC)

assay

≥98% (GC)

assay

≥99% (GC)

biological source

synthetic (organic)

biological source

synthetic

biological source

synthetic (organic)

biological source

synthetic (organic)

technique(s)

gas chromatography (GC): suitable

technique(s)

gas chromatography (GC): suitable, inhibition assay: suitable

technique(s)

gas chromatography (GC): suitable

technique(s)

gas chromatography (GC): suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

H2O: soluble 330 mg/mL, clear, colorless

solubility

H2O: 0.250 g/5mL

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

water: 50 mg/mL, clear, colorless

form

crystalline

form

crystalline

form

powder

form

solid


Still not finding the right product?

Explore all of our products under 2-Deoxy-D-glucose


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Rosemarie Ungricht et al.
The Journal of cell biology, 209(5), 687-703 (2015-06-10)
Newly synthesized membrane proteins are constantly sorted from the endoplasmic reticulum (ER) to various membranous compartments. How proteins specifically enrich at the inner nuclear membrane (INM) is not well understood. We have established a visual in vitro assay to measure
Alfredo J Ibáñez et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), 8790-8794 (2013-05-15)
Single-cell level measurements are necessary to characterize the intrinsic biological variability in a population of cells. In this study, we demonstrate that, with the microarrays for mass spectrometry platform, we are able to observe this variability. We monitor environmentally (2-deoxy-D-glucose)
Melissa A Burmeister et al.
American journal of physiology. Endocrinology and metabolism, 304(7), E677-E685 (2013-01-24)
Glucagon-like peptide-1 (GLP-1) suppresses food intake via activation of a central (i.e., brain) GLP-1 receptor (GLP-1R). Central AMP-activated protein kinase (AMPK) is a nutrient-sensitive regulator of food intake that is inhibited by anorectic signals. The anorectic effect elicited by hindbrain



Global Trade Item Number

SKUGTIN
D6134-5G04061833587300
D6134-250MG04061833587287
D6134-1G04061833587270
D6134-25G04061833587294

Questions

1–9 of 9 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. CAN THIS PRODUCT BE PREPARED IN WATER, AND FROZEN IN ALIQUOTS FOR LONG-TERM STORAGE?

    1 answer
    1. There is no information on the solution stability of aqueous solutions of this product. So it would be best to prepare fresh solutions as needed.

      Helpful?

  4. Can this product be prepared in DMSO, and frozen in aliquots for long-term storage?

    1 answer
    1. The chemical can be solubilized in DMSO (up to 190 mg/ml). Per literature reference, 1 M stock solutions of 2-deoxyglucose can be stored at -20 °C. Brandizzi, F., et al., Membrane Protein Transport between the Endoplasmic Reticulum and the Golgi in Tobacco Leaves Is Energy Dependent but Cytoskeleton Independent: Evidence from Selective Photobleaching. The Plant Cell, 14, 1293-1309 (2002). (Direct link to paper: http://www.plantcell.org/content/14/6/1293.full)

      Helpful?

  5. How can I dissolve 2-Deoxy-D-glucose, Product D6134?

    1 answer
    1. The product is soluble in water at 330 mg/mL.  It may also be soluble in DMSO.

      Helpful?

  6. Can solutions of 2-Deoxy-D-glucose, Product D6134, be stored?

    1 answer
    1. Sigma-Aldrich has no information on the solution stability of this product.  If information can't be found in the literature, it is best to prepare fresh solutions.

      Helpful?

  7. What are the differences between products D6134 and D8375 which are both 2-deoxy-D-glucose?

    1 answer
    1. Product D6134 is a further purification by crystallization of D8375.

      Helpful?

  8. What is the shelf life of 2-Deoxy-D-glucose, Product D6134?

    1 answer
    1. There is no recommended retest date given to released lots for this product. The product is guaranteed to be of good quality for one year from the time you receive it; this guarantee applies only to unopened bottles which have been stored at room temperature.

      Helpful?

  9. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters