Skip to Content
Merck

Skip To

D4693

Dispase® II

protease

Synonym(s):

Protease from Bacillus polymyxa

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
EC Number:
255-914-4
NACRES:
NA.54
MDL number:
Specific activity:
≥0.5 units/mg solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

lyophilized solid

specific activity

≥0.5 units/mg solid

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

solubility

10 mM NaAc (pH 7.5) and 5 mM CaAc: soluble

storage temp.

2-8°C

Application

The enzyme from Sigma has been used in developing a protocol for ex vivo culture of mouse embryonic mammary buds.[1] It has been used in the treatment of rat heart pieces during the isolation of mitochondria from rat heart.[2] It has also been used for the isolation of dental pulp stem cells (DPSCs) by enzymatic hydrolysis. These cells have been compared with DPSCs isolated by explant method to analyse their stem cell and differentiation properties.[3]
Dispase® II has been used for Fluorescence-Activated Cell Sorting (FACS).[4] It has also been used for separating visceral yolk sac layers.[5]
Suitable for use in preparation of single cell suspension for sequencing.

Biochem/physiol Actions

Dispase® II is a neutral protease that hydrolyzes the N-terminal peptide bonds of non-polar amino acid residues. It may be used for separating many tissues and cells grown in vitro. The enzyme is very gentle and does not damage cell membranes. It can also be used to prevent clumping in suspension cultures. This protease cleaves fibronectin and type IV collagen, but not laminin, type V collagen, serum albumin, or transferrin.[6] Dispase® II is specific for the cleavage of Leucine-Phenylalanine bonds. Ca2+, Mg2+, Mn2+, Fe2+, Fe3+ and Al3+ activate the enzyme. EDTA, EGTA, Hg2+ and other heavy metals inhibit the enzyme activity.[7] The enzyme contains 1g-atom of zinc per g-mol of purified enzyme. If this zinc component is removed by chelating agents such as EDTA or EGTA, an inactive apoenzyme is obtained. The enzyme is not inhibited by serum.

Physical form

lyophilized powder containing calcium acetate and milk sugar

Preparation Note

A 50 U/mL stock solution may be prepared by dissolving the powder in a buffer containing 10 mM NaAc (pH 7.5) and 5 mM CaAc. It should be stored at 4 °C.

Analysis Note

The enzyme can be initially dissolved in 50 mM Hepes/KOH pH 7.4, 150 mM NaCl at 10 mg/ml. Further dilutions should be made in the cell culture media most appropriate for the cells being dissociated. Dispase is a metalloproteinase and is activated by divalent cations including Ca2+ , Mg2+, Mn2+ and Fe2+.

Other Notes

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

Legal Information

Dispase is a registered trademark of Godo Shusei Co., Ltd.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D4818T9935C9891
technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable, single cell analysis: suitable

specific activity

≥0.5 units/mg solid

specific activity

≥10 unit/mg solid

specific activity

≥9,000 BAEE units/mg protein

specific activity

≥125 CDU/mg solid, 0.5-5.0 FALGPA units/mg solid

form

lyophilized solid

form

lyophilized solid

form

essentially salt-free, lyophilized powder

form

essentially salt-free, lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

10 mM NaAc (pH 7.5) and 5 mM CaAc: soluble

solubility

-

solubility

hydrochloric acid: soluble 1 mM

solubility

TESCA buffer (50 mM TES, 0.36 mM Calcium chloride, pH 7.4): soluble

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–8 of 8 Questions  
  1. Can I also dissolve the powder in 1x PBS?

    1 answer
    1. This material may be reconstituted in 1X PBS without Calcium and Magnesium. However, when this product is prepared as recommended, in 50 mM HEPES/KOH (pH 7.4) with 150 mM NaCl to 10 mg/mL, or in 10 mM sodium acetate (pH 7.5) with 5 mM calcium acetate, solutions are expected to be stable for 2 weeks at 2-8°C and up to 2 months when stored in aliquots at -20°C. Solution stability in PBS has not been evaluated. Various sources indicate solutions diluted in PBS are stable for approximately 3 days at 2–8 °C.

      Helpful?

  2. Can I aliquot in 10 mM sodium acetate (pH 7.5) + 5 mM calcium acetate, freeze once, and thaw as needed? I need small amounts from the same lot over several months, therefore I cannot store it at 2-8°degrees for only 2 weeks,

    1 answer
    1. Aliquots of this product prepared in 10 mM sodium acetate (pH 7.5) + 5 mM calcium acetate can be stored at -20 °C, where stability is known for up to 2 months.

      Helpful?

  3. How long can I store the powder at -80°C?

    1 answer
    1. In powder form, this product is quite stable and stored at 2-8°C. This product is not assigned an expiration or retest date. A warranty period of 1 year from the date of shipment applies. The long term stability of this material when stored at -20°C or -80°C has not been investigated. Prepared solutions are expected to be stable for 2 weeks at 2-8°C and up to 2 months when stored in aliquots at -20°C.

      Helpful?

  4. Could I dissolve in another work solution instead Hepes/KOH 50 mM pH 7,4, NaCl 150 mM? 

    1 answer
    1. Two approaches are available for reconstituting this lyophilized enzyme formulation. The first approach is for making a solution used in cell dissociation, and the second is a more generic method, such as for assaying Dispase activity:

      1. Dissolve in 50 mM Hepes/KOH pH 7.4, 150 mM NaCl at 10 mg/mL. Further dilutions should be made in the cell culture media most appropriate for the cells being dissociated.

      2. A 50 U/mL stock solution may be prepared by dissolving the powder in a buffer containing 10 mM sodium acetate (pH 7.5) and 5 mM calcium acetate.

      Helpful?

  5. how many units in 1 mg for the product D4693

    1 answer
    1. The enzymatic activity varies from lot to lot and is reported in the Certificate of Analysis. A brief survey of recent lots reveals an average activity of 1.2 u/mg solid. Please see the link below to review a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/d4693*product-documentation

      Helpful?

  6. Once dissolved into a stock solution, how long is it stable at 4 degrees C?

    1 answer
    1. The reconstituted stock solution can be stored at 2 to 8 °C for 2 weeks. Alternatively, it can be stored at - 20 deg C in aliquots to minimize freeze-thaw cycles where it should be stable for up to 2 months. The working solution, diluted in PBS from the stock solution, is stable for approximately 3 days at 2 to 8 °C.

      Helpful?

  7. Is Dispase II esteril for isolating cells from tissues for doing cell cultures? Thanks Monica

    1 answer
    1. This product has been used in the ex vivo culture of mouse embryonic mammary buds. Please see protocols in the published article: https://link.springer.com/article/10.1007/s10911-013-9288-2

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters