Skip to Content
Merck

Skip To

D1152

Dulbecco′s Modified Eagle′s Medium - high glucose

HEPES Modification, With 4500 mg/L glucose, L-glutamine, and 25 mM HEPES, without sodium bicarbonate and pyruvate, powder, suitable for cell culture

Synonym(s):

DME, DMEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

technique(s)

cell culture | mammalian: suitable

components

sodium pyruvate: no
NaHCO3: no
phenol red: yes
HEPES: 25 mM
L-glutamine: yes
glucose: high

shipped in

ambient

storage temp.

2-8°C

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Preparation Note

Formulated to contain 17.3 grams of powder per liter of medium.
Supplement with 3.7 g/L sodium bicarbonate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D7777D5648D6171
form

powder

form

powder

form

powder

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

components

sodium pyruvate: no
NaHCO3: no
phenol red: yes
HEPES: 25 mM
L-glutamine: yes
glucose: high

components

glucose: high
sodium pyruvate: yes
NaHCO3: no
HEPES: no
phenol red: yes
L-glutamine: yes

components

phenol red: yes
L-glutamine: yes
NaHCO3: no
HEPES: no
glucose: high
sodium pyruvate: no

components

sodium pyruvate: no
HEPES: 25 mM
glucose: high
NaHCO3: yes
L-glutamine: no
phenol red: yes

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

classic media, classical media, FBS, glucose enriched

classic media, classical media, FBS, glucose enriched

classic media, classical media, FBS, glucose enriched

classic media, classical media, serum supplement, FBS, HEPES, high glucose, pyridoxine


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Kiran Pandey et al.
Animal reproduction science, 181, 93-102 (2017-04-24)
Gonadotropin-releasing hormone receptors (GnRHRs) colocalize with insulin and glucocorticoid receptors in lipid rafts of the gonadotroph plasma membrane, where they facilitate downstream signaling. We recently found that orphan G-protein-coupled receptor (GPR)61 is expressed in the anterior pituitary (AP) of heifers
Jiping Sun et al.
Journal of cellular and molecular medicine, 22(10), 4840-4855 (2018-07-20)
BMSCs are important in replacement therapy of diabetic nephropathy (DN). MiR-124a exerts effect on the differentiation capability of pancreatic progenitor cells. The objective of this study was to explore the molecular mechanisms, the functions of miR-124a and bone marrow mesenchymal
Jian Xu et al.
Redox biology, 12, 198-207 (2017-03-05)
Physapubescin B, a steroidal compound extracted from the plant Physalis pubescens L. (Solanaceae), has been reported to possess anti-cancer potential, whereas the molecular mechanism remains elusive. In this study, we first demonstrated that physapubescin B induced autophagy in human cancer



Global Trade Item Number

SKUGTIN
D1152-10L04061838259226
D1152-50K04061834180180
D1152-10X1L04061838259233
D1152-BIBC04061826573556
D1152-50L04061833558775

Questions

Reviews

No rating value

Active Filters