Skip to Content
Merck

Skip To

CS0013

Red Protease Detection Kit

Fluorescent protease assay, high sensitivity

Synonym(s):

Fluorescent Protease Assay Kit, Protease Assay Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.51
UNSPSC Code:
12350000
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

packaging

pkg of 1 kit

shipped in

dry ice

storage temp.

−20°C

General description

The Red Protease Detection Kit uses TAMRA-casein as a convenient fluorescent substrate for detection of proteases in various samples.

Fluorescence excitation wavelength = 535 nm

Fluorescence emission wavelength = 595 nm

Application

Detection and quantitation of protease activity in samples. Useful for impurity testing.
Detection limit approximately 19 ng/ml, when using a 1 hour incubation with Trypsin as a standard.

Features and Benefits

  • Fluorescence is pH-independent
  • No separation or precipation steps are required
  • High sensitivity and signal-to-noise

Other Notes

CS0013A Trypsin (1 mg/mL): 100 μL

CS0013B Protease AssayBuffer × 2: 30 mL

CS0013C TAMRA-Casein substrate: 280 μL


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
CS0013-1KT04065269368434

Questions

Reviews

No rating value

Active Filters