Skip to Content
Merck

Skip To

CS0004

Nuclease micrococcal from Staphylococcus aureus

With reaction buffer and BSA solution

Synonym(s):

Nuclease micrococcal from Staphylococcus aureus, Endonuclease micrococcal, MNase, Micrococcal endonuclease

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77
EC Number:
MDL number:
Biological source:
Staphylococcus aureus
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Staphylococcus aureus

form

liquid

shipped in

wet ice

storage temp.

−20°C

General description

Nuclease Micrococcal (MNase) is the extracellular nuclease of Staphylococcus aureus. The enzyme is an endo-exonuclease .This Endo-exonuclease preferentially digests single-stranded nucleic acids. Thus, nuclease micrococcal activity results in the production of mononucleotides and oligonucleotides with terminal 3′-phosphates. Nuclease Micrococcal is also active against double-stranded DNA and RNA and all sequences will be ultimately cleaved.
Micrococcal Nuclease can be used for the digestion of chromatin to identify the location of nucleosomes

Application

  • Chromatin Immuno-Percipitation (ChIP) analysis
  • Nucleosome preparation
  • Removal of nucleic acids present in protein preparation
  • Reduction of viscosity of cell lysates during non-mechanical cell lysis preparation
  • RNA sequencing and transcriptomic profiling

Biochem/physiol Actions

Hydrolyzes 5′-phosphodiester bonds of DNA.

Other Notes

Note: One μmolar unit = approx. 85 A260 units, where an A260 unit is equivalent to a ΔA260 of 1.0 in 30 min at pH 8.8 at 37 °C in a 3.55 mL reaction volume. Light path = 1 cm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N3755N5386N5661
biological source

Staphylococcus aureus

biological source

Staphylococcus aureus

biological source

Staphylococcus aureus

biological source

Aspergillus sp. (A. oryzae)

form

liquid

form

lyophilized powder

form

powder

form

solution

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Lingli Long et al.
Journal for immunotherapy of cancer, 9(12) (2022-01-01)
Ovarian cancer (OvCa)-tumor-associated macrophages (TAMs) spheroids are abundantly present within ascites of high malignant patients. This study investigated the mutual interaction of OvCa cells and TAMs in the spheroids. Three-dimensional coculture system and transwell coculture system were created to mimic



Questions

Reviews

No rating value

Active Filters