Skip to Content
Merck

Skip To

C6682

Nunc® Lab-Tek® Chamber Slide system

2 wells, Permanox® slide, 4.2 cm2/well, sterile, 96/cs

Synonym(s):

cell culture chamber, chamber slides, microscope slides, Nunc 177429, microscopy slide system

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41122600
Material:
colorless , plastic (Permanox)
Sterility:
sterile
Packaging:
pack of 16, case of 96
W × l:
25 mm × 75 mm
Wells:
2
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

colorless , plastic (Permanox)

sterility

sterile

feature

CE marked

packaging

pack of 16, case of 96

manufacturer/tradename

Nunc 177429

W × L

25 mm × 75 mm

well area

4.2 cm2

wells

2

working volume

1.2-2.0 mL

suitability

suitable for (viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology)

General description

Grow, stain, and examine cells directly on the Nunc® Lab-Tek® Chamber Slide, which consists of a polystyrene media chamber attached to a microscopic slide. The slide surface is specially treated for adherent cell culture. Choose either a specialty washed soda lime glass slide or cell attachment optimized Permanox® plastic slide. Media chamber is easily removable without the need for a tool.

Application

  • Microscopy of in situ tissue culture
  • Viral and mycoplasma testing
  • Chromosome studies
  • Toxicity testing
  • Immunocytology

Features and Benefits

  • Cells grow on a standard microscope slide
  • Growth chamber can be removed when culturing is complete
  • Certified surface treatment ensures optimal contitions for cell attachment and growth
  • No cell transfer needed prior to visualisation/staining
  • Fits standard equipment
  • Saves time and reagents
  • Permanox™ does not interfere with fluorescent labels

Legal Information

Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Permanox is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C6932C7182C6807
W × L

25 mm × 75 mm

W × L

25 mm × 75 mm

W × L

-

W × L

-

manufacturer/tradename

Nunc 177429

manufacturer/tradename

Nunc 177437

manufacturer/tradename

Nunc 177445

manufacturer/tradename

Nunc 154526

material

colorless , plastic (Permanox)

material

colorless , plastic (Permanox)

material

-

material

colorless , polystyrene chamber

sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

wells

2

wells

4

wells

-

wells

4

packaging

pack of 16, case of 96

packaging

pack of 16, case of 96

packaging

pack of 16, case of 96

packaging

pack of 16 ea, case of 96 ea


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
C6682-1PAK04061834379911
C6682-1CS04061834373988