Skip to Content
Merck

Skip To

B7216

Bleomycin ready made solution

Cell culture tested, 10 mg/mL in H₂O

Synonym(s):

BLM, BLM A2, Blenoxane, Bleo, Bleomycin, Bleomycin A2, Blexane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.76
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Bleomycin ready made solution, Cell culture tested, 10 mg/mL in H2O

Quality Level

form

liquid

storage condition

protect from light

concentration

10 mg/mL in H2O

storage temp.

−20°C

General description

Bleomycin (BLM) is a glycopeptide antibiotic consist of 2 main isomers, BLM A2 and B2, obtained from Streptomyces verticillus. The main clinical application of Bleomycin is cancer treatment. Bleomycin causes oxidative degradation of RNA affecting G2 and M cell cycle phases. The anti-cancer mechanism of Bleomycin is mediated through iron chelation and generation of oxygen free radicals, leading to DNA cleavage and ultimately to cell death. Bleomycin hydrolase, the enzyme responsible for Bleomycin metabolism, is normally present at low levels in the lungs. Therefore, Bleomycin treatment may result in its accumulation in the lungs and can potentially cause lung injury due to lung cell death. The lung injury occurs through overproduction of reactive oxygen species causing pulmonary toxicity leading to fibrosis. The lung fibrosis by Bleomycin can be utilized for screening of antifibrotic agents.

Bleomycin has several important applications:
  • Bleomycin has been used to induce chronic lung disease in animal models as part of a research to find antifibrotic therapy for COVID-19 (SARS-CoV-2).
  • Bleomycin - induced pulmonary fibrosis in-vivo experiments performed in mice by dose administration of 0.25-2mg/Kg.
  • Bleomycin serves as a selective agent for gram-negative and gram-positive bacteria. Bleomycin cause DNA cleavage through formation of 1:1 complex with BLE protein. The resistance is obtained by encoding the ble gene of BLE protein from Streptoalloteichus hindustanus which prevents DNA cleavage.
  • Bleomycin can be used for selection in different types of cell lines.

Bleomycin suggested concentration range for cell selection is 10-100μg/mL. Hence, it is recommended to dilute the ready made solution 1:100-1,000 in media of choice.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML3001P4512M5353
Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

form

liquid

form

liquid

form

liquid

form

liquid

concentration

10 mg/mL in H2O

concentration

-

concentration

10 mg/mL in H2O

concentration

10 mg/mL in DMSO

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage condition

protect from light

storage condition

protect from light

storage condition

-

storage condition

protect from light


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Muta. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
B7216-1ML04065265040709

Questions

  1. What is the specific activity (U/mg) of the belomycin?

    1 answer
    1. The activity of this product is not determined. The material is prepared as a 10 mg/mL solution. However, historical data indicates the activity of the Bleomycin used to prepare this product may range from 1.5 - 2.0 units per milligram.

      Helpful?

Reviews

No rating value

Active Filters