Skip to Content
Merck

Skip To

B2434

Bleomycin sulfate from Streptomyces verticillus

BioXtra, crystalline

Bleomycin sulfate from Streptomyces verticillus
1 of 1 reviewers received a sample product or took part in a promotion

Synonym(s):

Blenoxane, Bleo, Blexane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
51111733
EC Number:
232-925-2
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioXtra

Quality Segment

form

crystalline

potency

1.5-2.0 units per mg

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 20 mg/mL, clear, colorless

cation traces

Cu: ≤0.10%

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, neoplastics

mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

[O-]S([O-])(=O)=O.C[C@@H](O)[C@@H](NC(=O)[C@@H](C)[C@H](O)[C@@H](C)NC(=O)[C@H](NC(=O)c1nc(nc(N)c1C)[C@H](CC(N)=O)NC[C@H](N)C(N)=O)[C@@H](O[C@@H]2O[C@@H](CO)[C@@H](O)[C@H](O)[C@@H]2O[C@@H]3O[C@H](CO)[C@@H](O)[C@@H](OC(N)=O)[C@@H]3O)c4c[nH]cn4)C(=O)NCCc5nc(cs5)-c6ncc(s6)C(=O)NCCC[S+](C)C.C[C@@H](O)[C@@H](NC(=O)[C@@H](C)[C@H](O)[C@@H](C)NC(=O)[C@H](NC(=O)c7nc(nc(N)c7C)[C@H](CC(N)=O)NC[C@H](N)C(N)=O)[C@@H](O[C@@H]8O[C@@H](CO)[C@@H](O)[C@H](O)[C@@H]8O[C@@H]9O[C@H](CO)[C@@H](O)[C@@H](OC(N)=O)[C@@H]9O)c%10c[nH]cn%10)C(=O)NCCc%11nc(cs%11)-c%12ncc(s%12)C(=O)NCCC[S+](C)C

InChI

1S/2C55H83N17O21S3.H2O4S/c2*1-20-33(69-46(72-44(20)58)25(12-31(57)76)64-13-24(56)45(59)82)50(86)71-35(41(26-14-61-19-66-26)91-54-43(39(80)37(78)28(16-73)90-54)92-53-40(81)42(93-55(60)88)38(79)29(17-74)89-53)51(87)67-22(3)36(77)21(2)47(83)70-34(23(4)75)49(85)63-10-8-32-68-27(18-94-32)52-65-15-30(95-52)48(84)62-9-7-11-96(5)6;1-5(2,3)4/h2*14-15,18-19,21-25,28-29,34-43,53-54,64,73-75,77-81H,7-13,16-17,56H2,1-6H3,(H13-,57,58,59,60,61,62,63,66,67,69,70,71,72,76,82,83,84,85,86,87,88);(H2,1,2,3,4)/t2*21-,22+,23+,24-,25-,28-,29+,34+,35+,36-,37+,38+,39-,40-,41-,42+,43-,53-,54-;/m00./s1

InChI key

OOXTWFJZZAJGKA-CNLAFNBISA-N

General description

Chemical structure: glycopeptide

Application

Bleomycin Sulfate is an antineoplastic antibiotic isolated from Streptomyces verticillus. It is recommended for use as a cell culture agent at 10-100 μg/mL.

Biochem/physiol Actions

Bleomycin sulfate binds to DNA, causes ssDNA scission at specific base sequences and inhibits DNA synthesis. This inhibitory action requires bleomycin to bind oxygen and a metal ion. It can also cleave RNA, to a lesser degree but more selectively. It acts as an inducer and regulator of apoptosis and inhibits tumor angiogenesis.

Preparation Note

The product is soluble in water in water at 20 mg/mL and is active for several days. A solution of 2 units/mL in 0.1 M neutral potassium phosphate should be used within 14 days. A solution is active for three months at 3 units/mL in normal saline.

Other Notes

Keep container tightly closed in a dry and well-ventilated place, strongly hygroscopic
This product is a mixture of glycopeptides antibiotics containing Bleomycin A2 (70%) and B2(30%). Bleomycins differ from each other in the terminal amine and show varying biological activity.

Disclaimer

This product should be stored at 2-8°C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
15361B5507B8416
antibiotic activity spectrum

Gram-negative bacteria, neoplastics, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, neoplastics

antibiotic activity spectrum

fungi

antibiotic activity spectrum

fungi

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

cell culture | mammalian: suitable

mode of action

DNA synthesis | interferes

mode of action

DNA synthesis | interferes

mode of action

DNA synthesis | interferes

mode of action

DNA synthesis | interferes

product line

BioXtra

product line

-

product line

-

product line

BioReagent

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

crystalline

form

powder or crystals

form

crystalline

form

powder or crystals


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Muta. 1B - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. What is the unit/mg for this product?

    1 answer
    1. This product has a potency specification of 1.5-2.0 units per mg. The exact value will be listed on the lot specific Certificate of Analysis. See the link below to review a sample Certificate of Analysis: https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/B2434-BULKSLCM6973.pdf

      Helpful?

  2. I can't find an expiration or use by date for the powder. Is it better to resuspend and freeze upon arrival or is it better to keep at 4deg until ready to resuspend and freeze?

    1 answer
    1. This product does not contain an expiration date or recommended retest date in its crystalline format. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment.

      This product should be placed in the proper storage temperature of 2-8°C upon arrival. Once reconstituted, the material should be stored at 2-8°C as well. For reconstitution instructions and stability, please see "Preparation Notes" section of the product page.

      Helpful?

Reviews

Active Filters

  1. Boston, MA
    • Review 1
    • Votes 0
    3 out of 5 stars.

    Needs to optimize product packaging (container)

    Could be beneficial if the vial is smaller or the product is in liquid form. The quantity is too little with respect to the size of the container, also the product (powder) sticks to the container

    Helpful?

    1. Response from Merck KGaA:

      Thank you for taking the time to leave a review! We appreciate your feedback and have passed on your concerns to our Product Management teams. Please do not hesitate to contact us at https://www.sigmaaldrich.com/support/customer-support and submit a Product Technical Inquiry for further assistance.