Skip to Content
Merck
All Photos(5)

Key Documents

A3263

Sigma-Aldrich

Alcohol Dehydrogenase from Saccharomyces cerevisiae

greener alternative

powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)

Synonym(s):

ADH1, ADH, Alcohol Dehydrogenase from yeast, Alcohol:NAD+ oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bakers yeast

form

powder

specific activity

≥300 units/mg protein

mol wt

~141,000 (four subunits)

purified by

crystallization

storage condition

(Tightly closed. Dry)

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

beige

optimum pH

8.6-9.0

suitability

suitable for recycling micro-assay of β-NAD and β-NADH

UniProt accession no.

greener alternative category

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Neuroscience

Yeast alcohol dehydrogenase 1 (ADH1) belongs to the family of zinc-containing alcohol dehydrogenases. It is a homotetramer with each subunit containing one catalytic domain and coenzyme-binding domain.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in fuel cell research. For more information see the article in biofiles.

Application

Alcohol Dehydrogenase from Saccharomyces cerevisiae has been used for the determination of NAD+ and NADH concentrations.

Biochem/physiol Actions

ADH (alcohol dehydrogenase) is one of the first enzymes to be isolated and purified. NAD+ is its coenzyme. Three isozymes of yeast ADH, that is, yeast alcohol dehydrogenase-1, 2 and 3 (YADH-1, -2, -3) have been identified. YADH-1 is expressed during anaerobic fermentation, YADH-2 is expressed in the cytoplasm and YADH-3 is localized to the mitochondria. A 141kDa tetramer containing 4 equal subunits. The active site of each subunit contains a zinc atom. Each active site also contains 2 reactive sulfhydryl groups and a histidine residue.

Isoelectric point: 5.4-5.8

Optimal pH: 8.6-9.0

Substrates: Yeast ADH is most active with ethanol and its activity decreases as the size of the alcohol increases or decreases. Branched chain alcohols and secondary alcohols also have very low activity.

KM (ethanol) = 2.1 × 10-2 M
KM (methanol = 1.3 × 10-1 M
KM (isopropanol) = 1.4 × 10-1 M

Inhibitors: Compounds that react with free sulfhydryls, including N-alkylmaleimides and iodoacetamide.
Zinc chelator inhibitors, including 1,10-phenanthroline,
8-hydroxyquinoline, 2,2′-dipyridyl, and thiourea.
Substrate analogue inhibitors, including β-NAD analogs, purine and pyrimidine derivatives, chloroethanol, and fluoroethanol.

Extinction Coefficient: E1% = 14.6 (water, 280 nm)
Yeast alcohol dehydrogenase 1 (YADH1) catalyzes the conversion of acetaldehyde to ethanol during glucose fermentation pathway. It is also implicated in the production of alcohol from amino acid breakdown via the Ehrlich pathway.

Specifications

The dried enzyme has been stored for several weeks in a vacuum desiccator with little loss in activity. According to experiments described by A. Kornberg,3 the enzyme can be stored in the frozen state and can be thawed repeatedly without marked loss of activity.

Unit Definition

One unit will convert 1.0 μmole of ethanol to acetaldehyde per min at pH 8.8 at 25 °C.

Physical form

Solids containing <2% citrate buffer salts

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nada J Habeichi et al.
Bioscience reports, 40(6) (2020-06-11)
The impact of cigarette smoking (CS) on kidney homeostasis in the presence of myocardial infarction (MI) in both males and females remains poorly elucidated. C57BL6/J mice were exposed to 2 weeks of CS prior to MI induction followed by 1
Zsofia Komary et al.
Biochimica et biophysica acta, 1777(7-8), 800-807 (2008-06-05)
Release of H(2)O(2) in response to Ca(2+) loads (1-100 microM) was investigated using Amplex red fluorescent assay in isolated guinea-pig brain mitochondria respiring on glutamate plus malate or succinate. In mitochondria challenged with Ca(2+) (10 microM), in the absence of
The role of zinc in alcohol dehydrogenase. V. The effect of metal-binding agents on thestructure of the yeast alcohol dehydrogenase molecule.
J H KAGI et al.
The Journal of biological chemistry, 235, 3188-3192 (1960-11-01)
N C Jerez et al.
Journal of animal science, 81(4), 997-1003 (2003-05-02)
The objective of this experiment was to determine the effect of prerigor injection of several glycolytic inhibitors on pH, color, tenderness, and related traits of low-value beef cuts. The semimembranosus, triceps brachii, and supraspinatus muscles from each of 10 steer
Sexual dimorphism in acute myocardial infarction-induced acute kidney injury: cardiorenal deteriorating effects of ovariectomy in premenopausal female mice
Nada J Habeichi, et al.
Clinical Science (London, England : 1979), 137(1), 47-63 (2023)

Protocols

To measure alcohol dehydrogenase activity, this assay uses β-nicotinamide adenine dinucleotide phosphate and a continuous spectrophotometric rate determination at 340 nm.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service