Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
form
solid
fluorescence
λex 492 nm; λem 517 nm in 0.1 M Tris pH 8.0
storage temp.
−20°C
Analysis Note
unconjugated dye ≤5% of total fluorescence
Legal Information
NeutrAvidin is a registered trademark of Thermo Fisher Scientific, Inc.
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.