Skip to Content
Merck

Skip To

920P

Trilogy®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous solution

Quality Level

packaging

pkg of 1 L ready to use (920P-09), pkg of 1 gal ready to use (920P-10), pkg of 200 mL 20x (920P-06), pkg of 200 mL ready to use (920P-05), pkg of 240 mL 100x (920P-07), pkg of 50 mL 20x (920P-04)

technique(s)

immunohistochemistry: suitable

shipped in

ambient

General description

Trilogy is the ultimate solution for your lab. Unlike other EDTA solutions, Trilogy is a near neutral pH solution, making it less harmful on tissue, while still exhibiting the strong unmasking abilities of a conventional EDTA solution. Trilogy′s revolutionary formula combines deparaffinization, rehydration, and unmasking, all in one simple step! Trilogy allows for standardization of the pretreatment procedure, which in turn leads to more consistent and reliable results.

Physical form

Solution containing pretreatment reagents

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Trilogy is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
922P1.990911.99019
technique(s)

immunohistochemistry: suitable

technique(s)

immunohistochemistry: suitable

technique(s)

-

technique(s)

-

Quality Level

500

Quality Level

100, 500

Quality Level

300

Quality Level

300

form

aqueous solution

form

aqueous solution

form

-

form

-

packaging

pkg of 1 L ready to use (920P-09), pkg of 200 mL 20x (920P-06), pkg of 240 mL 100x (920P-07), pkg of 1 gal ready to use (920P-10), pkg of 50 mL 20x (920P-04), pkg of 200 mL ready to use (920P-05)

packaging

pkg of 1 L ready-to-use (922P-09), pkg of 200 mL concentrated 20x (922P-06), pkg of 200 mL ready-to-use (922P-05), pkg of 240 mL concentrated 100x (922P-07), pkg of 50 mL concentrated 20x (922P-04)

packaging

-

packaging

-

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Pamungkas Bagus Satriyo et al.
Journal of clinical medicine, 8(2) (2019-01-30)
Cancer stem cells (CSCs) promote tumor progression and distant metastasis in breast cancer. Cadherin 11 (CDH11) is overexpressed in invasive breast cancer cells and implicated in distant bone metastases in several cancers. The WNT signalling pathway regulates CSC activity. Growing
Allison R Pine et al.
Cancer discovery, 10(7), 964-979 (2020-04-08)
Glioblastoma (GBM), an incurable tumor, remains difficult to model and more importantly to treat due to its genetic/epigenetic heterogeneity and plasticity across cellular states. The ability of current tumor models to recapitulate the cellular states found in primary tumors remains
Amanda Linkous et al.
Cell reports, 26(12), 3203-3211 (2019-03-21)
The prognosis of patients with glioblastoma (GBM) remains dismal, with a median survival of approximately 15 months. Current preclinical GBM models are limited by the lack of a "normal" human microenvironment and the inability of many tumor cell lines to accurately reproduce



Global Trade Item Number

SKUGTIN
920P-060841683144922
920P-040841683144908
920P-070841683106715
920P-050841683144915
920P-090841683106739
920P-100841683106722

Questions

1–7 of 7 Questions  
  1. Is the Trilogy-920P-09 we purchased 20X? Do we need to dilute it before use? So we don’t need to perform deparaffinization and hydration with xylene and gradient alcohol? Also, does the solution contain Triton and Tween?

    1 answer
    1. Item # 920P-10 is 1 gallon of ready to use Trilogy and 920P-09 is one liter of ready to use Trilogy.  No dilution is needed for either of these part numbers.

      Helpful?

  2. Can Trilogy be re-used, and is it used with the PT Module or other automation?

    1 answer
    1. There is no official data to support the reusability of the product.

      Helpful?

  3. Can you provide information regarding how Trilogy was made?

    1 answer
    1. The EDTA-based buffer is a concentrate that needs to be diluted as 20X.

      Helpful?

  4. What is the pH of 1X Trilogy reagent

    1 answer
    1. The concentrate product Trilogy has a variable pH ranging from 7.8 to 8.8.

      Helpful?

  5. What are the troubleshooting methods for the product Trilogy 200 mL concentrate (20x) (catalogue number 920P-06)? There was a nonspecific 3,3'-diaminobenzidine (DAB) signals in IHC experiments on paraffin-embedded kidney sections using the 1x Trilogy buffer and a rice cooker for the heating steps. Is there a suggestion for optimizing the antigen retrieval steps of Trilogy to achieve a milder unmasking effect?

    1 answer
    1. The issue could be related to fixation. A protocol for use in a steamer involves using 50mls to make 1 liter of solution, with 30 minutes in the steamer and 30 minutes in hot rinse with agitation and no cool down. Subsequently, rinsing with wash buffer is crucial, requiring several rinses for cleaner staining. An alternative option is to use Trilogy to deparaffinize and rehydrate, leaving it in the steamer for 60 minutes before proceeding to rinsing.

      Helpful?

  6. Is Trilogy a fully aqueous solution or does it contain any organic solvent? How does the deparaffinization process using Trilogy compare to the process using organic solvents and a series of alcohol concentrations? Can Trilogy be reused, and is there a protocol available for using it? Additionally, is it necessary to extend the time of deparaffinization when using large FFPE sections of human brain with Trilogy?

    1 answer
    1. Trilogy is an EDTA-based buffer with a variable pH of 7.8 to 8.8. It also contains some detergents/surfactants. An automatic pressure cooker is recommended for use as it tends to give the most consistent results. It may take some experimentation to work with a pressure cooker for the brain specimens to ensure that you do not experience any adhesion issues. Another heat source option might be a steamer, but this will take longer (45 minutes to an hour) for the process to be complete for unmasking. Also, the second container used for rinsing during the initial process can be refrigerated for use as the starting buffer for the next batch of slides.

      Helpful?

  7. What are the differences between Trilogy and Declere, apart from Trilogy containing Triton X in its original formulation?

    1 answer
    1. Trilogy is EDTA-based and has a higher pH than Declere, which is citrate buffer-based. Trilogy is recommended for the majority of all antibodies, while Declere is used when visualizing degrees of expression, such as with Her2Nu where the use of citrate buffer is recommended.

      Helpful?

Reviews

No rating value

Active Filters