Skip to Content
Merck

Skip To

90C2

Leukocyte Naphthol AS-D Chloroacetate (Specific Esterase) Kit

Synonym(s):

Leder Stain

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
12352106
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Select reagents packaged in gelatin capsules

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

dilution

(for histology)

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

Esterases are ubiquitous in nature and encompass a series of different enzymes acting upon select substrates. With the 90C2 kit, Naphthol AS-D Chloroacetate is enzymatically hydrolyzed by "specific esterase, " liberating a free naphthol compound. This then couples with a diazonium salt, forming highly colored deposits at sites of enzyme activity. This enzyme is usually considered specific for cells of granulocytic lineage. May be used to detect neutrophils in peripheral blood, bone marrow or tissue sections which have been paraffin embedded.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
91CHT25A911
application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

wet ice

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

description

Select reagents packaged in gelatin capsules

description

Kit formulated with all liquid reagents

description

-

description

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C


Still not finding the right product?


Kit Components Only

Product No.
Description

  • Dimethyl formamide (9010) 25 mL

  • Fast Corinth V Salt (9015) 10 capsules

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Acid Hematoxylin Solution 100 mL
    SDS

  • Naphthol AS-D Chloroacetate 10 x 20
    SDS

  • TRIZMAL™ 6.3 Buffer Concentrate, pH 6.3(concentrate), 0.2 M 50 mL
    SDS

signalword

Danger

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B - STOT RE 2 Oral

target_organs

Central nervous system



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



J Heijmans et al.
Oncogene, 32(9), 1202-1206 (2012-04-04)
Development of colon cancer is a multistep process that is regulated by intrinsic and extrinsic cellular signals. Extrinsic factors include molecular patterns that are derived from either pathogens (PAMPs) or cellular damage (DAMPs). These molecules can promote tumourigenesis by activation
John Zagorski et al.
Journal of immunology (Baltimore, Md. : 1950), 179(11), 7820-7826 (2007-11-21)
Right ventricular (RV) dysfunction is a strong risk factor for poor clinical outcome following pulmonary embolism (PE), the third most prevalent cardiovascular disease. Previous studies in our laboratory demonstrated that RV failure during PE is mediated, in part, by neutrophil-dependant
Chengjie Ma et al.
Molecular medicine reports, 22(1), 483-493 (2020-04-23)
Inactivation of the Hippo pathway protects the myocardium from cardiac ischemic injury. MicroRNAs (miRs) have been reported to play pivotal roles in the progression of myocardial infarction (MI). The present study examined whether miR‑93 could promote angiogenesis and attenuate remodeling



Global Trade Item Number

SKUGTIN
90C2-1KT04061833647806

Questions

  1. Regarding the 90C2-1KT kit: Are all kit components included? How many slides can I stain with this 90C2 Kit?

    1 answer
    1. The kit components required to stain slides are included in the kit, which is designed for 10 uses. Each use typically requires approximately 50 mL of staining reagent, sufficient to stain from 1 to 13 or 14 slides. This means the kit can stain a minimum of 10 slides or a maximum of 130-140 slides.

      Helpful?

Reviews

No rating value

Active Filters