Skip to Content
Merck

Skip To

90278

Triethanolamine

BioUltra, ≥99.5% (GC)

Synonym(s):

2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
UNSPSC Code:
12161700
NACRES:
NA.25
PubChem Substance ID:
EC Number:
203-049-8
Beilstein/REAXYS Number:
1699263
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

5.14 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 20 °C)

product line

BioUltra

assay

≥99.5% (GC)

form

liquid, viscous liquid

autoignition temp.

600 °F

expl. lim.

8.5 %

impurities

insoluble matter, passes filter test, ≤0.1% ethanolamine (GC), ≤0.2% water, ≤0.3% diethanolamine (GC)

ign. residue

≤0.1% (as SO4)

refractive index

n20/D 1.485

pH

10.5-11.5 (25 °C, 1 M in H2O)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.)

mp

17.9-21 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

1.124 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg, sulfate (SO42-): ≤20 mg/kg

SMILES string

OCCN(CCO)CCO

cation traces

Al: ≤2 mg/kg, As: ≤0.1 mg/kg, Ba: ≤1 mg/kg, Bi: ≤2 mg/kg, Ca: ≤5 mg/kg, Cd: ≤1 mg/kg, Co: ≤1 mg/kg, Cr: ≤1 mg/kg, Cu: ≤1 mg/kg, Fe: ≤1 mg/kg, K: ≤20 mg/kg, Li: ≤1 mg/kg, Mg: ≤1 mg/kg, Mn: ≤1 mg/kg, Mo: ≤1 mg/kg, Na: ≤20 mg/kg, Ni: ≤1 mg/kg, Pb: ≤1 mg/kg, Sr: ≤1 mg/kg, Zn: ≤1 mg/kg

absorption

cut-off at 270 nm in H2O at 1 M

λ

1 M in H2O

UV absorption

λ: 280 nm Amax: 0.1

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

Application

Triethanolamine (TEA), an emulsifier, surfactant and capping agent, may be used to study its toxicity profile and mechanisms. TEA may be used in various drug delivery constructs to improve drug solubility. TEA may be used to cap CdTe nanoparticles.

Other Notes

Buffer in recommended method for alanine amionopeptidase in urine.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
337299027916303
assay

≥99.5% (GC)

assay

≥99% (GC)

assay

≥99.0% (GC)

assay

≥99% (GC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

product line

BioUltra

product line

-

product line

-

product line

-

form

liquid, viscous liquid

form

viscous liquid

form

liquid, viscous liquid

form

-

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

soluble 149 g/L at 20 °C (completely)

solubility

H2O: 1 M, clear, colorless

solubility

-

density

1.124 g/mL at 25 °C (lit.)

density

1.124 g/mL at 25 °C (lit.)

density

1.124 g/mL at 25 °C (lit.)

density

1.124 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Triethanolamine


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

354.2 °F - closed cup

flash_point_c

179 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



H Mattenheimer et al.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie, 26(10), 635-644 (1988-10-01)
A method is recommended for the measurement of the catalytic concentration of alanine aminopeptidase in the urine of man, rat, and dog, using L-alanine-4-nitroanilide as substrate. In currently used methods, substrate concentrations between 1.7 and 2.0 mmol/l are used. Kinetic
Liviu Ungur et al.
Journal of the American Chemical Society, 134(45), 18554-18557 (2012-11-01)
A toroidal magnetic moment in the absence of conventional total magnetic moment is observed in a {Dy(6)} ring. The reason for the net toroidal arrangement of the local magnetic moments is the high symmetry of the complex in combination with
Stuart K Langley et al.
Dalton transactions (Cambridge, England : 2003), 41(32), 9789-9796 (2012-07-11)
The synthesis, structures and magnetic properties of two new mixed-valence heptanuclear manganese clusters are described. Both complexes utilize triethanolamine (teaH(3)) as a bridging ligand, displaying near planar, disc-like metal topologies and are of formulae [Mn(II)(4)Mn(IV)(3)(tea)(teaH(2))(3)(peolH)(4)](BF(4))(2)·solv (1) and [Mn(II)(4)Mn(III)(3)F(3)(tea)(teaH)(teaH(2))(2)(piv)(4)(Hpiv)(chp)(3)]·0.5MeCN (2). Compound



Global Trade Item Number

SKUGTIN
90278-500ML04061833246597
90278-100ML04061833246580

Questions

Reviews

No rating value

Active Filters