Skip to Content
Merck

Skip To

86250

α-Amylase from Aspergillus oryzae

greener alternative

powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)

Synonym(s):

Taka-Amylase A, Taka-Diastase from Aspergillus oryzae, 1,4-α-D-Glucan-glucanohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
232-588-1
NACRES:
NA.54
MDL number:
EC Number:
Specific activity:
~1.5 U/mg (~0.2 U acc. to Willstätter)
Biological source:
Aspergillus sp. (A. oryzae)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Aspergillus sp. (A. oryzae)

Quality Segment

form

powder

specific activity

~1.5 U/mg (~0.2 U acc. to Willstätter)

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch hydrolysis research. For more information see the article in biofiles.

Other Notes

1 U corresponds to the amount of enzyme which liberates 1 μmol maltose per minute at pH 6.0 and 25 °C (starch acc. to Zulkowsky, Cat. No. 85642, as substrate).
Molecular weight; Product distribution in the hydrolysis of amylose; Application in (selective) hydrolysis/condensation of glycosidic bonds; Suitable for use in thiamine determination.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9857A822010065
specific activity

~1.5 U/mg (~0.2 U acc. to Willstätter)

specific activity

≥150 units/mg protein (biuret)

specific activity

≥800 FAU/g

specific activity

~30 U/mg

biological source

Aspergillus sp. (A. oryzae)

biological source

-

biological source

Aspergillus sp. (A. oryzae)

biological source

-

form

powder

form

powder

form

aqueous solution

form

powder

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

greener alternative category

Enabling,

greener alternative category

Enabling,

greener alternative category

Enabling,

greener alternative category

-


Still not finding the right product?

Explore all of our products under α-Amylase from Aspergillus oryzae


pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
86250-100G04061835553136
86250-500G04061833040812

Questions

  1. Can you kindly indicate the optimum pH and temperature for this enzyme?

    1 answer
    1. As indicated above under ‘Unit Definition’ the enzyme’s activity is tested at pH 6.0 and 25 °C (starch acc. to Zulkowsky, Cat. No. 85642, as substrate).

      Optimal pH, pH and temperature stability of the enzyme, as well as effects of chemicals on its activity are addressed in the article below:
      Production of Amylase in Liquid Culture by a Strain of Aspergillus oryzae
      Kundu, A.K. and Das S.
      Applied Microbiology, 19(4), 598-603 (1970)

      Optimal pH and temperature ranges can also be found in the Brenda database:
      https://www.brenda-enzymes.org/enzyme.php?ecno=3.2.1.1

      Helpful?

Reviews

No rating value

Active Filters