Skip to Content
Merck

Skip To

84797

Poly(ethylene glycol)

greener alternative

BioUltra, 2,000

Synonym(s):

Polyethylene glycol, PEG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352104
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

BioUltra

form

powder

mol wt

Mr 1900-2200

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

insoluble matter:, passes filter test, ≤0.001% peroxides (as H2O2):

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

mp

50-53 °C

solubility

H2O: 50 mg/mL at 25 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: ≤0.1, λ: 280 nm Amax: ≤0.04

greener alternative category

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. Polyethylene glycol (PEG) is an eco-friendly, biodegradable polymer widely used in pharmaceuticals and cosmetics. Its non-toxic nature and versatility make it a sustainable choice, derived from renewable resources, contributing to greener product formulations. Click here for more information.
Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system, which is required for the purification of biological materials. PEG also interacts with cell membrane, thereby allowing cell fusion. It exhibitsstealth effect, prolonged blood circulation and excellent biocompatibility. PEG has an ability to elicit hypersensitivity reactions via stimulating complement pathway.

Application

Poly(ethylene glycol) has been used:
  • as a constituent in the preparation of liquid desiccant solutions
  • as a precipitating agent and additive in protein crystallization
  • as a representative biocompatible polymer to decipher its interactions with cellular system at molecular level[1]


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
812558951088276
form

powder

form

solid

form

solid

form

powder

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mol wt

Mr 1900-2200

mol wt

Mr 5000-7000

mol wt

Mr 7000-9000

mol wt

Mr 3000-3700

cation traces

Al: ≤5 mg/kg, Ba: ≤5 mg/kg, Ca: ≤10 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, K: ≤200 mg/kg, Mg: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Sr: ≤5 mg/kg, As: ≤0.1 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Mn: ≤5 mg/kg, Pb: ≤5 mg/kg, Bi: ≤5 mg/kg, Li: ≤5 mg/kg, Zn: ≤5 mg/kg, Cr: ≤5 mg/kg, Na: ≤200 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤5 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

impurities

insoluble matter:, passes filter test, ≤0.001% peroxides (as H2O2):

impurities

insoluble matter, passes filter test, ≤20 mg/kg peroxides (as H2O2)

impurities

insoluble matter:, passes filter test, ≤0.001% peroxides (as H2O2):

impurities

insoluble matters, passes filter test

mp

50-53 °C

mp

-

mp

61-64 °C

mp

-


Still not finding the right product?

Explore all of our products under Poly(ethylene glycol)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

281.5 °F - closed cup

flash_point_c

138.6 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Influence of osmotic drying with an aqueous poly (ethylene glycol) liquid desiccant on alumina objects gelcast with gelatin
Hammel EC, et al.
Ceramics International, 43(18), 16443-16450 (2017)
Poly(Ethylene Glycol) Chemistry: Biotechnical and Biomedical Applications (2013)
Exploring carbon nanomaterial diversity for nucleation of protein crystals
Govada L, et al.
Scientific Reports, 6(18), 20053-20053 (2016)



Global Trade Item Number

SKUGTIN
142905-250MG04061838734532
84797-1KG-F04061833008126
84797-250G-F04061833008133

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters