The solubility of the item is chloroform/methanol (9:1): 20 mg/mL.
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
soybean
Quality Segment
assay
≥98% (TLC)
form
powder
composition
16:0, ~35% , 18:0, ~8% , 18:1, ~7% , 18:2, ~40% , 18:3, ~5%
solubility
chloroform/methanol (9:1): 20 mg/mL, clear, colorless to very faintly yellow
functional group
phospholipid
lipid type
phosphoglycerides
shipped in
wet ice
storage temp.
−20°C
SMILES string
[Na+].CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)O[C@@H]1[C@H](O)[C@H](O)[C@@H](O)[C@H](O)[C@H]1O)OC(=O)CCCCCCCCCCCCCCCCC
InChI
1S/C45H87O13P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(46)55-35-37(36-56-59(53,54)58-45-43(51)41(49)40(48)42(50)44(45)52)57-39(47)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h37,40-45,48-52H,3-36H2,1-2H3,(H,53,54);/q;+1/p-1/t37-,40-,41-,42+,43-,44-,45-;/m1./s1
InChI key
OSYCDGBQPGKSOV-GJCFFWOHSA-M
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (TLC) | assay ≥98.0% (TLC) | assay ≥97.0% | assay >99% (TLC) |
| biological source soybean | biological source soybean | biological source soybean | biological source - |
| functional group phospholipid | functional group phospholipid | functional group phospholipid | functional group - |
| solubility chloroform/methanol (9:1): 20 mg/mL, clear, colorless to very faintly yellow | solubility - | solubility - | solubility - |
| shipped in wet ice | shipped in ambient | shipped in - | shipped in dry ice |
| form powder | form liquid | form solution | form powder |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Hallo, wir haben das Produkt bei uns im Labor. Können Sie mir bitte etwas zur Löslichkeit sagen. --> welches Lösungsmittel, Konzentration ?
1 answer-
Helpful?
-



