Skip to Content
Merck

Skip To

75351

Oligomycin A

≥99% (HPLC)

Oligomycin A

Synonym(s):

MCH 32

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C45H74O11
CAS Number:
Molecular Weight:
791.06
UNSPSC Code:
51102829
NACRES:
NA.76
PubChem Substance ID:
EC Number:
209-437-3
Beilstein/REAXYS Number:
5702132
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: soluble

antibiotic activity spectrum

fungi

mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

CCC1CCC2OC3(CC[C@@H](C)C(C[C@H](C)O)O3)[C@H](C)C(OC(=O)\C=C\[C@@H](C)[C@H](O)[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@H](C)C(=O)[C@@](C)(O)[C@H](O)[C@@H](C)C\C=C\C=C\1)[C@H]2C

InChI

1S/C45H74O11/c1-12-34-17-15-13-14-16-27(4)42(51)44(11,53)43(52)32(9)40(50)31(8)39(49)30(7)38(48)26(3)18-21-37(47)54-41-29(6)35(20-19-34)55-45(33(41)10)23-22-25(2)36(56-45)24-28(5)46/h13-15,17-18,21,25-36,38,40-42,46,48,50-51,53H,12,16,19-20,22-24H2,1-11H3/b14-13+,17-15+,21-18+/t25-,26-,27+,28+,29+,30-,31+,32+,33-,34?,35?,36?,38+,40-,41?,42-,44+,45?/m1/s1

InChI key

MNULEGDCPYONBU-VVXVDZGXSA-N

General description

Chemical structure: macrolide

Application

Oligomycin was used to inhibit sucrose synthesis and 14C translocation in sunflower leaves, to study antibiotic resistance, and to study carotid chemoreceptor responses to O2 and CO2 in the cat. Inhibits the growth of Rhodotorula glutinis, Aspergillus niger and other molds.[1]

Biochem/physiol Actions

Oligomycin blocks oxidative phosphorylation and the electron transport chain by inhibiting membrane bound mitochondrial ATP synthase (ATPase) and proton channel (pump, FO subunit) blocker.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place. Keep in a dry place.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
O487649545559983
antibiotic activity spectrum

fungi

antibiotic activity spectrum

fungi

antibiotic activity spectrum

-

antibiotic activity spectrum

Gram-positive bacteria

mode of action

enzyme | inhibits

mode of action

enzyme | inhibits

mode of action

-

mode of action

protein synthesis | interferes

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

powder

form

powder

form

powder

form

powder

assay

≥99% (HPLC)

assay

≥90% total oligomycins basis (HPLC)

assay

≥90% (mixture of A, B, and C isomers, HPLC)

assay

≥90% (HPLC)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Oligomycin A


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
75351-5MG04061835541751

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. For how long does the product remain stable once dissolved, aliquoted and stored at -20°C?

    1 answer
    1. Storage of stock solutions at -20°C is stable for 1 month.

      Helpful?

  4. Does this product contain thimerosal or any mercury-containing preservatives?

    1 answer
    1. This is a pure product, not containing any preservative or thimerosal.

      Helpful?

  5. What's it's solubility in dmso

    1 answer
    1. This product is not tested for solubility in DMSO, however other Oligomycin products have been found to be soluble at in DMSO at 50 mg/ml.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review