Skip to Content
Merck
All Photos(1)

Documents

71725

Sigma-Aldrich

Sodium dodecyl sulfate

BioUltra, for molecular biology, ≥99.0% (GC)

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161900
eCl@ss:
39093306
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

description

anionic

product line

BioUltra

Assay

≥99.0% (GC)

form

powder

mol wt

micellar avg mol wt 18,000

aggregation number

62

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

loss

≤3% loss on drying, 20 °C (HV)

CMC

7-10 mM (20-25°C)

mp

204-207 °C (lit.)

transition temp

cloud point >100 °C

solubility

H2O: 0.1 M at 20 °C, clear, colorless

density

0.455 g/cm3

anion traces

chloride (Cl-): ≤200 mg/kg
phosphate (PO43-): ≤1 mg/kg

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.04
λ: 280 nm Amax: 0.02

HLB

40

suitability

suitable for electrophoresis

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium dodecyl sulfate (SDS), also known as sodium lauryl sulfate, is a versatile anionic surfactant with a broad range of applications in molecular biology, biochemistry, and various industries. Its amphiphilic nature, characterized by a hydrophobic hydrocarbon tail and a hydrophilic sulfate head group, makes it an effective detergent capable of solubilizing and denaturing proteins, lipids, and other hydrophobic molecules.

In protein biochemistry, SDS plays a pivotal role in protein extraction and SDS-PAGE (sodium dodecyl sulfate-polyacrylamide gel electrophoresis). Its ability to denature proteins by disrupting their secondary and tertiary structures allows for their uniform migration during gel electrophoresis, enabling the separation and analysis of proteins based on their molecular weight. This technique has become an indispensable tool for identifying and characterizing proteins in biological samples.

Beyond its role in protein analysis, SDS also finds utility in nucleic acid extraction and various hybridization techniques. Its detergent properties aid in the lysis of cells and the release of nucleic acids, facilitating their subsequent purification and analysis. Additionally, SDS is employed in hybridization techniques to promote the annealing of complementary nucleic acid strands, enabling the detection and analysis of specific DNA or RNA sequences.

Application

Sodium dodecyl sulfate has been used:
  • as an ionic surfactant in polystyrene (PS) particle suspension for surfactant-based experiments
  • in phosphate-buffered saline as a component of lysis buffer to homogenize myometrium and leiomyoma tissue
  • as a component of lysis buffer to dissolve formazans
  • as a component of high-salt nuclear extraction buffer

Anionic detergent

Biochem/physiol Actions

Sodium dodecyl sulfate (SDS) serves as an important detergent in the structural biology field and protein folding/unfolding studies. The sulfate groups and alkyl chains of SDS can strongly bind to the positively charged and the hydrophobic residues of proteins. SDS induces the formation of α-helices in a protein structure. At high concentrations, SDS stimulates cooperative unfolding in various proteins such as bovine serum albumin (BSA), hen egg-white lysozyme, -lactalbumin, and -lactoglobulin structures.

Features and Benefits

  • Suitable for molecular biology and biochemical research
  • BioUltra grade for your sensitive applications
  • Free from DNase, RNase, NICKase and protease
  • Tested for trace levels of Anions and Cations
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications

  • Undergoes hydrolysis at high temperatures, especially in acidic medium

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of in vitro translation products.
R Jagus
Methods in enzymology, 152, 296-304 (1987-01-01)
Shane Stegeman et al.
Endocrine-related cancer, 22(2), 265-276 (2015-02-12)
The oncogene MDM4, also known as MDMX or HDMX, contributes to cancer susceptibility and progression through its capacity to negatively regulate a range of genes with tumour-suppressive functions. As part of a recent genome-wide association study it was determined that
Kazuya Ishikawa et al.
Nature communications, 5, 5430-5430 (2014-11-13)
Pathogen effector proteins are delivered to host cells to suppress plant immunity. However, the mechanisms by which effector proteins function are largely unknown. Here we show that expression of XopP(Xoo), an effector of rice pathogen Xanthomonas oryzae pv. oryzae, in
Hannah M Kaneb et al.
Human molecular genetics, 24(5), 1363-1373 (2014-10-26)
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disorder characterized by the selective death of motor neurons. Causative mutations in the global RNA-processing proteins TDP-43 and FUS among others, as well as their aggregation in ALS patients, have identified defects
Akiko Kitamura et al.
The Journal of experimental medicine, 211(12), 2385-2396 (2014-11-12)
Autoinflammatory syndromes cause sterile inflammation in the absence of any signs of autoimmune responses. Familial cold autoinflammatory syndrome (FCAS) is characterized by intermittent episodes of rash, arthralgia, and fever after exposure to cold stimuli. We have identified a missense mutation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service