Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-773-7
MDL number:
Specific activity:
≥50 U/mg
Biological source:
Porcine liver
Pricing and availability is not currently available.
biological source
Porcine liver
Quality Segment
form
lyophilized solid, powder
quality
lyophilized
specific activity
≥50 U/mg
mol wt
Mr ~162000
color
slightly beige
storage temp.
−20°C
General description
Porcine liver esterase (PLE) is localized in the endoplasmic reticulum (ER).[1]
Application
Pig liver esterase is commonly used for kinetic resolutions and assymetric synthesis in organic chemistry[2].
Esterase from porcine liver has been used as a negative control in fluorescence measurement studies.[3]
Porcine liver esterase is used to catalyze the hydrolysis of pentaacetyl catechin and epicatechin for use in pharmaceutical and industrial applications.
Pig liver esterase is commonly used for kinetic resolutions and assymetric synthesis in organic chemistry.
Pig liver esterase is commonly used for kinetic resolutions and assymetric synthesis in organic chemistry.
Biochem/physiol Actions
Esterase acts on water-soluble carboxyl esters containing short chain fatty acids. Its functionality is attributed to the catalytic triad of Ser, His and Asp/Glu.
Other Notes
1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol ethyl valerate (CAt. No. 30784) per minute at pH 8.0 and 25°C
contains 1,4-dithioerythritol
Disclaimer
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| biological source Porcine liver | biological source - | biological source - | biological source - |
| specific activity ≥50 U/mg | specific activity ≥30 units/mg protein | specific activity ≥15 units/mg solid | specific activity ≥0.2 U/mg |
| form lyophilized solid, powder | form lyophilized powder | form lyophilized powder | form lyophilized |
| mol wt Mr ~162000 | mol wt 60 kDa | mol wt 168 kDa | mol wt - |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| color slightly beige | color - | color - | color - |
Still not finding the right product?
Explore all of our products under Esterase from porcine liver
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




