Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.84
UNSPSC Code:
12161503
Pricing and availability is not currently available.
General description
The P450 Human Induction 100 Assay Kit provides a complete protocol, instrument data acquisition methods, processing methods and stable isotope labeled synthetic peptide standards for the analysis of four human CYP450 isoforms (1A2, 2B6, 3A4, and 3A5) for induction studies using LC/MS/MS. Kit contains enough reagents for 100 assays. Must order Trypsin TPCK (500 μg) and Peptide C18 Column separately.
Analysis Note
To view the Protocol for the CYP450 Protein Assay - Human Induction Kit, please visit this Protocol link.
To view the Quick Reference Card for the CYP450 Protein Assay - Human Induction Kit, please visit this Quick Reference Card link.
A Technical Note on the use of the CYP450 Protein Assay - Human Induction Kit is available at this Tech Note link.
To view the Quick Reference Card for the CYP450 Protein Assay - Human Induction Kit, please visit this Quick Reference Card link.
A Technical Note on the use of the CYP450 Protein Assay - Human Induction Kit is available at this Tech Note link.
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) LC/MS: suitable | technique(s) LC/MS: suitable | technique(s) LC/MS: suitable | technique(s) ELISA: suitable, capture ELISA: suitable |
| storage temp. −20°C | storage temp. - | storage temp. - | storage temp. −20°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level - |
signalword
Danger
Storage Class
3 - Flammable liquids
wgk
WGK 3
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Central nervous system
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 4445252-1KT | 04061833648483 |





