Skip to Content
Merck

Skip To

380B

Sudan Black B Staining System

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
12352106
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

Expiry date on the label.

Quality Level

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

General description

Intended for the histochemical demonstration of neutrophil granules in blood or bone marrow films.

Biochem/physiol Actions

Following fixation, blood or bone marrow films are immersed in a buffered Sudan Black B solution. After rinsing, slides are counterstained with Gill hematoxylin, and cells are examined microscopically for the presence of blue-black discrete granulation. Granulocytic precursors demonstrate groups of coarse blue-black deposits. Other hematopoietic cells are typically negative.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
199664S4131395B
application(s)

hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

hematology
histology

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

wet ice

IVD

for in vitro diagnostic use

IVD

-

IVD

-

IVD

for in vitro diagnostic use

shelf life

Expiry date on the label.

shelf life

-

shelf life

-

shelf life

Expiry date on the label.

storage temp.

2-8°C

storage temp.

room temp

storage temp.

room temp

storage temp.

2-8°C

Quality Level

500

Quality Level

200

Quality Level

200

Quality Level

500


Kit Components Only

Product No.
Description

  • Sudan Black B Staining Reagent (3801) 200 mL

  • Hematoxylin Solution, Gill No. 3 (GHS3) 100 mL

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Glutaraldehyde solution, for in vitro diagnostic use 75 mL
    SDS

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 2 - STOT SE 3

target_organs

Eyes,Central nervous system, Kidney, Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

flash_point_f

69.1 °F

flash_point_c

20.6 °C

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



F Zhao et al.
Leukemia, 32(1), 149-158 (2017-06-20)
Interferon regulatory factor (IRF)-8 is a critical transcription factor involved in the pathogenesis of myeloid neoplasia. However, the underlying mechanisms in vivo are not well known. Investigation of irf8-mutant zebrafish in this study indicated that Irf8 is evolutionarily conserved as
Yuelan Hou et al.
Scientific reports, 6, 36853-36853 (2016-11-12)
Emergency granulopoiesis occurs in response to severe microbial infection. However, whether and how other blood components, particularly monocytes/macrophages and their progenitors, including hematopoietic stem/progenitor cells (HSPCs), participate in the process and the underlying molecular mechanisms remain unknown. In this study
Ana-Maria Chiorcea-Paquim et al.
Analytical chemistry, 87(12), 6141-6149 (2015-05-12)
The interactions of the Tetrahymena telomeric repeat sequence d(TG4T) and the polyguanylic acid (poly(G)) sequence with the quadruplex-targeting triazole-linked acridine ligand GL15 were investigated using atomic force microscopy (AFM) at a highly oriented pyrolytic graphite and voltammetry at a glassy



Global Trade Item Number

SKUGTIN
380B-1KT04061831960860

Questions

  1. what is the molar weight of these product ? (g/mol)

    1 answer
    1. This product is a kit item that includes multiple components. The Sudan Black B reagent is a 0.18% solution. The molecular weight of this dye is 456.54 g/mol. Please see the link below to review the kit technical bulletin for additional information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/202/528/380.pdf

      Helpful?

Reviews

No rating value

Active Filters