Skip to Content
Merck

Skip To

13368

1-Deoxy-D-xylulose-5-phosphate sodium salt

≥99.0% (TLC)

Synonym(s):

DXP sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H11O7P · xNa+
CAS Number:
Molecular Weight:
214.11 (free acid basis)
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Beilstein/REAXYS Number:
8367371
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥99.0% (TLC)

form

powder

optical activity

[α]/D 37.0±3.0°, c = 0.1 in 0.1 M HCl

color

white

storage temp.

−20°C

SMILES string

[P](=O)(OC[C@@H](O)[C@H](O)C(=O)C)(O)O

InChI

1S/C5H11O7P/c1-3(6)5(8)4(7)2-12-13(9,10)11/h4-5,7-8H,2H2,1H3,(H2,9,10,11)/t4-,5-/m1/s1

InChI key

AJPADPZSRRUGHI-RFZPGFLSSA-N

Application

1-Deoxy-D-xylulose-5-phosphate is used as a substrate for the identification, differentiation and characterization of procaryotic 1-Deoxy-D-xylulose-5-phosphate reductoisomerase(s) (Dxr) which catalyze the first committed step of the nonmevalonate pathway (NMP) for isoprenoid biosynthesis.

Biochem/physiol Actions

Metabolite of the non-mevalonate pathway, generally found in prokaryotes, as precursor to isoprenoids as well asnon-isoprenoids like vitamins. As this pathway is not present in humans, it is of interest for the development of bacterium-specific drugs in the search for treatments of infectious diseases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



B M Lange et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(5), 2100-2104 (1998-04-16)
Isopentenyl diphosphate, the common precursor of all isoprenoids, has been widely assumed to be synthesized by the acetate/mevalonate pathway in all organisms. However, based on in vivo feeding experiments, isopentenyl diphosphate formation in several eubacteria, a green alga, and plant
W Eisenreich et al.
Chemistry & biology, 5(9), R221-R233 (1998-09-30)
Recent studies have uncovered the existence of an alternative, non-mevalonate pathway for the formation of isopentenyl pyrophosphate and dimethylallyl pyrophosphate, the two building blocks of terpene biosynthesis.
Y Boucher et al.
Molecular microbiology, 37(4), 703-716 (2000-09-06)
Lateral gene transfer (LGT) is a major force in microbial genome evolution. Here, we present an overview of lateral transfers affecting genes involved in isopentenyl diphosphate (IPP) synthesis. Two alternative metabolic pathways can synthesize this universal precursor of isoprenoids, the



Global Trade Item Number

SKUGTIN
13368-5MG04061838729590
13368-1MG04061838729583

Questions

Reviews

No rating value

Active Filters