Skip to Content
Merck

Skip To

PPB002

Tris Buffered Saline

powder, pH 8.0, pHast Pack, with 0.05% Tween® 20

Synonym(s):

Tris Buffered Saline, with Tween™ 20, pH 8.0, TBS with 0.05% Tween-20, TBS-T, TBST

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41105323
NACRES:
NA.21
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tris Buffered Saline, with 0.05% Tween® 20, pH 8.0, powder, pHast Pack

product line

pHast Pack

form

powder

storage condition

(Tightly closed. Dry.)

impurities

≤5 ppm Heavy Metals (lead)

color

white

pH

7.8—8.2

solubility

soluble, clear to hazy, colorless

cation traces

Fe: ≤10 ppm

foreign activity

DNase, none detected, Endonuclease , Exonuclease, none detected, NICKase, none detected, Protease, none detected, RNase, none detected

storage temp.

room temp

General description

Tris Buffered Saline with 0.05% Tween® 20 or TBST has a low concentration of detergent with a slightly basic pH of 8.0. It is a commonly used washing buffer alternative to PBS-T in many immunodetection techniques. It decreases non-specific bindings from the western blot membrane and microtiter plate walls.

Application

Suitable for use as a wash buffer in:
  • Western blot
  • ELISA
  • Immunohistochemistry

Features and Benefits

  • Ready-to-use buffer requires no measurments or pH adjustments
  • Saves time, money and effort to get your assays started fast
  • Physical, chemical, and biological tests for quality: free of DNase, RNase, Protease, and Nickase
  • Qualified for use in gel electrophoreis and other molecular biology assays

Packaging

Foil pouches

Preparation Note

Contents of one pouch, when dissolved in 500 mL of distilled or deionized water, will yield a 1× solution containing 50 mM Tris, 138 mM NaCl, 2.7 mM KCl; 0.05%Tween 20 (w/v), pH 8.0 at 25 °C. Certified to be DNAse, RNAse, Protease, and Nickase free.
Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

TWEEN is a registered trademark of Croda International PLC
pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PPB001T9039PPB011
product line

pHast Pack

product line

pHast Pack

product line

-

product line

pHast Pack

form

powder

form

powder

form

powder

form

powder

solubility

soluble, clear to hazy, colorless

solubility

soluble, clear, colorless

solubility

-

solubility

soluble, clear, colorless

storage condition

(Tightly closed. Dry.)

storage condition

(Tightly closed. Dry)

storage condition

-

storage condition

(Tightly closed. Dry)

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

room temp

pH

7.8—8.2

pH

8.0

pH

-

pH

7.6


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Questions

Reviews

No rating value

Active Filters