Skip to Content
Merck

Skip To

FPPMCC3

Flash Point Standard - Pensky Martens

certified reference material, ASTM D 93 Proc. A (116.0 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

according to ASTM® D93

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

petroleum
pharmaceutical

format

neat

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Pensky Martens Flash Point Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
FPPMCC4FPPMCC5FPPMCC6
format

neat

format

neat

format

neat

format

neat

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

petroleum
pharmaceutical

application(s)

petroleum
pharmaceutical

application(s)

petroleum
pharmaceutical

application(s)

petroleum
pharmaceutical

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


Still not finding the right product?


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

168.8 °F

flash_point_c

76 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
FPPMCC3-3X80ML04061838189639

Questions

Reviews

No rating value

Active Filters