Skip to Content
Merck

Skip To

75482

Gram′s decolorizer solution

for Gram staining

Synonym(s):

Acetone ethanol mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12171500
NACRES:
MA.02
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

technique(s)

microbe id | staining: suitable

refractive index

n20/D 1.361

density

0.80 g/mL at 20 °C

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

General description

Gram staining is one of the most significant staining techniques in microbiology. Gram′s decolorizer solution is a mixture of ethanol and acetone.

Application

Gram′s decolorizer solution is used in Gram staining procedure, after the iodine step to remove the crystal violet dye. The decolorizer dissolves the lipid layer from the gram-negative cells.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
946357773087794
technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

application(s)

diagnostic assay manufacturing
hematology
histology

form

liquid

form

liquid

form

-

form

liquid

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

room temp

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

density

0.80 g/mL at 20 °C

density

0.986 g/mL at 20 °C

density

-

density

0.999 g/mL at 20 °C


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

flash_point_f

68.0 °F

flash_point_c

20 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters