Skip to Content
Merck

Skip To

13404

Sodium aluminate

technical grade, powder

Synonym(s):

Aluminum sodium oxide, Sodium aluminum oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
NaAlO2
CAS Number:
Molecular Weight:
81.97
NACRES:
NA.22
UNSPSC Code:
12161600
EC Number:
234-391-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sodium aluminate, technical

grade

technical

Quality Segment

form

powder

reaction suitability

core: aluminum, reagent type: catalyst

impurities

≤0.05% Fe (as Fe2O3)

cation traces

Al (Al2O3): 50-56%, Na (as Na2O): 37-45%

SMILES string

[Na+].O=[Al-]=O

InChI

1S/Al.Na.2O/q-1;+1;;

InChI key

IYJYQHRNMMNLRH-UHFFFAOYSA-N

General description

Sodium aluminate is used in the synthesis of industrial catalyst mesoporous alumina and various aluminum trihydroxide polymorphs.

Application

Sodium aluminate can be used as a catalyst:
  • In the transesterification of glycerol with dimethyl carbonate to synthesize glycerol carbonate.
  • In the isomerization of D-glucose into D-fructose.
  • To synthesize biodiesel from Jatropha oil via transesterification.
  • To convert alcohols into the corresponding unsymmetrical carbonate esters via carboxymethylation by using dimethyl carbonate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
401307451584515884
reaction suitability

core: aluminum, reagent type: catalyst

reaction suitability

core: titanium, reagent type: catalyst

reaction suitability

core: sodium, reagent type: catalyst

reaction suitability

core: aluminum, reagent type: catalyst

form

powder

form

solid

form

powder

form

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

cation traces

Al (Al2O3): 50-56%, Na (as Na2O): 37-45%

cation traces

-

cation traces

-

cation traces

-

impurities

≤0.05% Fe (as Fe2O3)

impurities

-

impurities

≤150.0 ppm Trace Metal Analysis

impurities

-

grade

technical

grade

-

grade

-

grade

-


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1A

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Cleaner synthesis of mesoporous alumina from sodium aluminate solution
Zhong, Li and Zhang, et al.
Green Chemistry, 13, 2525-2530 (2011)
Rienacker, R.; Graefe, J.
Angewandte Chemie (International Edition in English), 24, 320-320 (1985)
Synthesis of Uniformly Spherical Bayerite from a Sodium Aluminate Solution Reacted with Sodium Bicarbonate
You, Shaowei and Li, et al.
Industrial & Engineering Chemistry Research, 52, 12710-12716 (2013)



Global Trade Item Number

SKUGTIN
326356-5G04061833606230
13404-50KG-R04061833544471
13404-2.5KG-R04061835561421
13404-5KG-R04061838729798

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Are the reported concentrations for 13404 Sodium aluminate in mole percent or wt%?

    1 answer
    1. The percentages reported on the Certificate of Analysis (CoA) for this product are in weight %.

      Helpful?

Reviews

No rating value

Active Filters