Skip to Content
Merck

Skip To

08511

Curcumin

analytical standard

Synonym(s):

(E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Diferuloylmethane, Diferulylmethane, Natural Yellow 3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
[HOC6H3(OCH3)CH=CHCO]2CH2
CAS Number:
Molecular Weight:
368.38
UNSPSC Code:
85151701
NACRES:
NA.24
E Number:
E100
PubChem Substance ID:
EC Number:
207-280-5
MDL number:
Colour Index Number:
75300
Beilstein/REAXYS Number:
2306965
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

13 (vs air)

assay

≥98.0% (HPLC)

form

powder

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)c(OC)c2)ccc1O

InChI

1S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-12,24-25H,13H2,1-2H3/b7-3+,8-4+

InChI key

VFLDPWHFBUODDF-FCXRPNKRSA-N

General description

Curcumin is an anti-inflammatory agent extracted from the roots of Curcuma longa Linn.

Application

Curcumin may be used as a reference standard for the determination of curcumin in rat plasma by high-performance liquid chromatography method.

Biochem/physiol Actions

A natural phenolic compound responsible for the yellow color of turmeric. Potent anti-tumor agent that acts against highly diverse tumor-specific pathways. Anti-inflammatory and anti-oxidant. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cyclooxygenase (COX-2) and 5-lipoxygenase (5-LOX).
A natural phenolic compound. Potent anti-tumor agent having anti-inflammatory and anti-oxidant properties. Curcumin has been cited as a potential chemopreventive agent, in addition to its chemotherapeutic activity. Induces apoptosis in cancer cells and inhibits phorbol ester-induced protein kinase C (PKC) activity. Reported to inhibit production of inflammatory cytokines by peripheral blood monocytes and alveolar macrophages. Potent inhibitor of EGFR tyrosine kinase and IκB kinase. Inhibits inducible nitric oxide synthase (iNOS), cycloxygenase and lipoxygenase. Easily penetrates into the cytoplasm of cells, accumulating in membranous structures such as plasma membrane, endoplasmic reticulum and nuclear envelope.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: curcuma

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2209C1386C7727
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

-

format

neat

format

-

format

-

format

-

assay

≥98.0% (HPLC)

assay

-

assay

-

assay

≥80% (Curcumin), ≥94% (curcuminoid content)

form

powder

form

solid

form

powder

form

powder

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Curcumin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A rapid and simple HPLC method for the determination of curcumin in rat plasma: assay development, validation and application to a pharmacokinetic study of curcumin liposome.
Li J, et al.
Biomedical Chromatography, 23(11), 1201-1207 (2009)
Oxygen radical scavenging activity of curcumin.
Kunchandy E and Rao MNA
International Journal of Pharmaceutics, 58(3), 237-240 (1990)
Phase I clinical trial of curcumin, a chemopreventive agent, in patients with highrisk or pre-malignant lesions.
Hsieh CY.
Anticancer Research, 21, 2895-2900 (2001)



Global Trade Item Number

SKUGTIN
261939-1G04061838118912
261939-5G04061838118929
08511-10MG04061838652508

Questions

  1. Good morning, do you know the solubility of curcumin in 70% ethanol and absolute ethanol? Thank you in advance.

    1 answer
    1. Curcumin is generally more soluble in pure ethanol. Ethanol's ability to dissolve curcumin is enhanced due to non-polar characteristics, which can effectively interact with the non-polar regions of curcumin. While curcumin is soluble in 70% ethanol, solubility is typically lower than in 100% ethanol.

      Helpful?

Reviews

No rating value

Active Filters