Skip to Content
Merck

Skip To

FIBRP-RO

Roche

Fibronectin (pure)

from human plasma, >95% (SDS-PAGE), lyophilized (clear, colorless solution after reconstitution)

Synonym(s):

Fibronectin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
MDL number:
NACRES:
NA.54
Biological source:
human plasma
Form:
lyophilized (clear, colorless solution after reconstitution)
Technique(s):
cell culture | mammalian: suitable
Mol wt:
440 kDa
Assay:
>95% (SDS-PAGE)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human plasma

sterility

non-sterile; 0.2 μm filtered

assay

>95% (SDS-PAGE)

form

lyophilized (clear, colorless solution after reconstitution)

mol wt

440 kDa

packaging

pkg of 1 mg (11051407001), pkg of 5 mg (11080938001)

manufacturer/tradename

Roche

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... FN1(2335)

General description

Fibronectin is responsible for the cellular interactions with the extracellular matrix. It is involved in cell adhesion, migration, growth and differentiation. It is a soluble component of plasma and other body fluids. In addition, it is also part of the insoluble extracellular matrix.[1]

Application

Fibronectin promotes the attachment and subsequent spreading of many cells. It is used for the coating of culture dishes.[2] It has been used for the cell adhesion assay.[3][4]

Biochem/physiol Actions

Active on most mammalian cells
Biological activity: Tested for the promotion of adherence of 3T3 (NR6) cells (XTT cleavage).

Physical form

Lyophilizate containing 1.126 mg glycine and 0.058 mg sodium chloride per mg fibronectin

Preparation Note

Working concentration: 5μg/cm2 for the coating of cell culture vessels
Storage conditions (working solution): -15 to -25 °C
The reconstituted solution is stable at -15 to -25 °C.
Note: Store the reconstituted solution in aliquots at -15 to -25 °C.
Avoid repeated freezing and thawing.
Add 1 or 5 ml sterile water, respectively, to yield a final concentration of 1 mg/ml. Incubate 30 to 60 minutes at 37 °C to dissolve. Do not agitate.
Upon reconstitution, the solution may contain a small amount of insoluble aggregated material. These aggregates are a phenomenon inherent to fibronectin and do not influence product performance.

Other Notes

For life science research only. Not for use in diagnostic procedures.
Human plasma fibronectin consists of two similar polypeptide chains ( α-, β-chain, each of 220 kDa), connected by two disulfide bridges.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
10838039001F2733MAB88916-C
biological source

human plasma

biological source

human plasma

biological source

human

biological source

mouse

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

ELISA: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, inhibition assay: suitable, western blot: suitable

sterility

non-sterile; 0.2 μm filtered

sterility

non-sterile; 0.2 μm filtered

sterility

-

sterility

-

assay

>95% (SDS-PAGE)

assay

95% (SDS-PAGE)

assay

-

assay

-

form

lyophilized (clear, colorless solution after reconstitution)

form

lyophilized (clear, colorless solution after reconstitution)

form

liquid

form

-

mol wt

440 kDa

mol wt

440 kDa

mol wt

-

mol wt

-


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Anindita Dutta et al.
International journal of clinical and experimental pathology, 3(3), 288-302 (2010-03-13)
The interaction of cells with adhesion proteins in the extracellular matrix (ECM) provides signals which affect the morphology, motility, gene expression and survival of adherent cells. In the present communication we cultured K562 cells in presence of fibronectin to study
Roberta Bulla et al.
Nature communications, 7, 10346-10346 (2016-02-03)
Complement C1q is the activator of the classical pathway. However, it is now recognized that C1q can exert functions unrelated to complement activation. Here we show that C1q, but not C4, is expressed in the stroma and vascular endothelium of
Fibronectin at a glance.
Roumen Pankov et al.
Journal of cell science, 115(Pt 20), 3861-3863 (2002-09-24)



Global Trade Item Number

SKUGTIN
1108093800104061838698292
1105140700104061838697493