Likely not, due to pore size/possible fouling/chemical compatibility. Refer to our guide, "Chemical compatibility of filter components": https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/302/855/membrane-chemical-compatibility-ds2532en-ms.pdf
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Segment
material
polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing
product line
Stericup®
feature
hydrophilic
packaging
pack of 12 ea
greener alternative product characteristics
Waste Prevention
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
parameter
max. temp. at 45 °C
description
2.4 mL (hold-up volume)
technique(s)
sterile filtration: suitable
H
225 mm
diam.
11.3 cm
filter capacity
1000 mL
filter diam.
73 mm
filtration area
40 cm2
matrix
Express PLUS™
pore size
0.22 μm pore size
fitting
inlet thread(s), 38 mm (1 1/2 in.) thread
greener alternative category
shipped in
ambient
storage temp.
no temp limit
General description
The Stericup® E and Steritop® E sterile filters are designed as a Greener Alternative for vacuum sterile filtration that significantly reduces plastic and packaging.
This Stericup® E is a 38mm thread collar which uses a 0.22 μm pore size polyethersulfone (PES) membrane filter unit combined with a 1000 mL receiver flask and cap for processing and storage. The 38 mm thread directly connects to our media bottles and majority of other standard commercial media bottles.
Use only glass or plastic bottles designed for vacuum applications.
Application
- Tissue culture media +/- additives
- Buffers
- Biological solutions
Features and Benefits
- The filter collar disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
- Bold product identification on the filter collar plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
- Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
- Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
- Click seal cap rests on the side to avoid risk of contamination.
- Receiver bottles are stackable to save storage space.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| pore size 0.22 μm pore size | pore size 0.22 μm pore size | pore size 0.22 μm pore size | pore size 0.22 μm pore size |
| filter capacity 1000 mL | filter capacity 500 mL | filter capacity 500 mL | filter capacity 1000 mL |
| material polyethersulfone membrane, polystyrene funnel(s), polystyrene bottle, polystyrene housing | material polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing | material polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing | material polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing |
| packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea |
| fitting inlet thread(s), 38 mm (1 1/2 in.) thread | fitting inlet thread(s), 45 mm thread | fitting inlet thread(s), 38 mm (1 1/2 in.) thread | fitting inlet thread(s), 45 mm thread |
| feature hydrophilic | feature hydrophilic | feature hydrophilic | feature hydrophilic |
Still not finding the right product?
Explore all of our products under Stericup® E-GP Funnel-less Sterile Vacuum Filtration System
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SEGPU1138 | 04061841074250 |
-
Can we filter oil-type liquids with Stericup® filters?
Can we filter oil-type liquids with Stericup® filters?
1 answer-
Helpful?
-
-
Which type of pump should be used with Stericup® and Steritop® vacuum filters?
Which type of pump should be used with Stericup® and Steritop® vacuum filters?
1 answer-
We recommend our Chemical Duty Pump: https://www.sigmaaldrich.com/product/mm/wp6111560
Helpful?
-
-
What is the maximum vacuum pressure for filtration?
1 answer-
700 mm Hg (27.5” Hg, 0.9 bar) differential vacuum at 25 °C (77 °F)
Helpful?
-
-
Can Stericup® and Steritop® filters be reused?
1 answer-
All Stericup® and Steritop® filters were developed for single use, disposable processing.
Helpful?
-
-
What tubing can be used with Stericup® and Steritop® vacuum filters?
What tubing can be used with Stericup® and Steritop® vacuum filters?
1 answer-
Stericup® and Steritop® filters are fitted with a 1/4” tube adapter. We recommend 3/16 in. (4.8 mm) ID silicone tubing to fit this tube adapter: https://www.sigmaaldrich.com/product/mm/xx7100004
Helpful?
-
-
Can Stericup® and Steritop® filters be used to prepare solutions that will be administered to patients?
1 answer-
No, all Stericup® and Steritop® devices are Research Use Only.
Helpful?
-
-
The User Guide says “Use only glass or plastic bottles designed for vacuum applications”. Can you specify which bottles are compatible?
1 answer-
Steritop® Quick Release bottle top units require a vacuum-rated receiver bottle with either a GL33 or GL45 mm closure thread as specified. Steritop® E bottle top units require a vacuum-rated receiver bottle with a GL45 mm closure thread. Bottles no larger than 2 L should be used.
Glass bottles, GL33:
• Wheaton media bottles (glass), 500 mL (Product Z364886)
• Wheaton media bottles (glass), 1 L (Product Z364894)
Glass bottles, GL45
• Corning® glass media storage bottle, 125 mL (Product CLS1399125)
• Corning® glass media storage bottle, 250 mL (Product CLS1399250)
• Corning® glass media storage bottle, 500 mL (Product CLS1399500)
• Corning® glass media storage bottle, 1 L (Product CLS13991L)
• Pyrex® round media storage bottle, 500 mL (Product CLS1395500)
• Pyrex® round media storage bottle, 1 L (Product CLS13951L)
• KIMBLE® KIMAX® Media bottle, 250 mL (Product DWK14395250)
• KIMBLE® KIMAX® Media bottle, 500 mL (Product DWK14395500)
• KIMBLE® KIMAX® Media bottle, 1 L (Product DWK143951000)
Plastic (pre-sterile) bottles, GL45:
• Stericup® Quick Release Receiver flask, 250 mL (Product S200B02RE)
• Stericup® Quick Release Receiver flask, 500 mL (Product S200B05RE)
• Stericup® Quick Release Receiver flask, 1000 mL (Product S200B10RE)Helpful?
-
-
Can Stericup® bottles be placed inside freezers for long-term storage of media?
1 answer-
You can successfully freeze and store many aqueous solutions (such as culture media) in Stericup® bottles at temperatures up to -20 °C (-4 °F). It is strongly recommended that you run a sample stability trial under your actual storage conditions prior to using Stericup® bottles for frozen storage.
Helpful?
-
-
What is the difference between classic Stericup® filters and Stericup® “Quick Release” filters?
1 answer-
Stericup® Quick Release, or “QR”, filters feature a connection between funnel filter and receiver bottle that requires only a quarter-turn release to facilitate funnel removal. These filters come fully engaged – there is no need to tighten before use. The receiver bottle cap has flanges on the top surface to improve grip, as well as a positive lock stop to ensure a leak-proof seal with the receiver bottle. Other improvements include a large, frosted writing surface on the receiver bottle and a lighter cap color to facilitate labelling, more prominent volume markings on the filtration funnel and receiver bottle, and a product label with a peel-off lot record strip on the bag for easy viewing in storage.
Helpful?
-
-
What Bacterial Endotoxins Test is used?
1 answer-
The test method used complies with USP 85 and EP 2.6.14 to ensure the absence of endotoxin in our products.
Helpful?
-



