This product contains lithium dodecyl sulfate with pH at 8.4, but does not include a reducing agent. Either Dithiothrietol (DDT) or beta-mercaptoethanol (BME) may be used. The recommended final concentration is 100 mM DTT or 25 mM BME. Please see the link below to access the mPAGE™ Bis-Tris Precast SDS-PAGE Gels User Guide, page 7:_x000D_
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/384/513/20344106w-ms.pdf
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
form
liquid
packaging
vial of 10 mL
General description
- Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Application
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form solution | form - |
| packaging vial of 10 mL | packaging vial of 250 mL | packaging - | packaging pkg of 1 pack |
Still not finding the right product?
Explore all of our products under mPAGE® 4X LDS Sample Buffer
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MPSB-10ML | 04054839761690 |
-
What is the composition of the loading buffer. Does it contain any reducing agents?
1 answer-
Helpful?
-
-
Can Laemmli buffer be used instead of the sample buffer?
1 answer-
Laemmli buffer is commonly used with Tris-Glycine gels, while mPAGE gels use a different chemistry (Bis-Tris) and require LDS buffer. Therefore, it is recommended to use the LDS buffer, which can be prepared using the formula provided in the mPAGE user guide.
Helpful?
-
-
Est ce que ce tampon contient déjà un agent réducteur tel que le DTT ou le béta mercapto ethanol ?
1 answer-
This product requires the addition of a reducing agent. Either Dithiothrietol (DDT) or beta-mercaptoethanol (BME) may be used. The recommended final concentration is 100 mM DTT or 25 mM BME. Please see the link below to access the mPAGE™ Bis-Tris Precast SDS-PAGE Gels User Guide, page 7:
https://www.emdmillipore.com/US/en/product/mPAGE-8-160-0-Bis-Tris-Precast-Gel-10x8-cm-12-well,MM_NF-MP81G12#documentationHelpful?
-




