Skip to Content
Merck

Skip To

A100-M

NovaSeptum® Sealing

NovaSeptum® Crimping Tool

Synonym(s):

Manual crimping tool for Novaseptum® pinch-pipes

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

eCl@ss:
36101790
UNSPSC Code:
41104105
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

sterility

non-sterile

product line

NovaSeptum®

application(s)

sterile sampling

compatibility

for use with NovaSeptum® tube disconnection pinch pipes. NS6-15-05PK100

Legal Information

NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A104-MNM0043NM0044
application(s)

sterile sampling

application(s)

sterile sampling

application(s)

-

application(s)

-

product line

NovaSeptum®

product line

NovaSeptum®

product line

NovaSeal

product line

NovaSeal

compatibility

for use with NovaSeptum® tube disconnection pinch pipes. NS6-15-05PK100

compatibility

-

compatibility

for use with NovaSeal pinch pipes

compatibility

for use with NovaSeal pinch pipes

Quality Level

SP2

Quality Level

SP2

Quality Level

100

Quality Level

100

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile


Still not finding the right product?

Explore all of our products under NovaSeptum® Sealing




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Questions

Reviews

No rating value

Active Filters