Skip to Content
Merck

Skip To

8.00181

Acrylic acid

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

Acrylic acid, 2-Propenoic acid, Acroleic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH2=CHCOOH
CAS Number:
Molecular Weight:
72.06
UNSPSC Code:
12352106
EC Index Number:
201-177-9
NACRES:
NA.22
MDL number:
eCl@ss:
39021317
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

3.8 hPa ( 20 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

390 °C (DIN 51794)

potency

>192 mg/kg LD50, oral (Rat), >290 mg/kg LD50, skin (Rabbit)

expl. lim.

3.9-19.8 % (v/v)

pH

2.1 (20 °C, 72.06 g/L in H2O)

bp

141 °C/1013 hPa

mp

13 °C

transition temp

flash point 46 °C

solubility

1000 g/L

density

1.05 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[O-]C(=O)C=C.[H+]

InChI

1S/C3H4O2/c1-2-3(4)5/h2H,1H2,(H,4,5)

InChI key

NIXOWILDQLNWCW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.048 - 1.050
Identity (IR): passes test
The material is stabilised with hydroquinone monomethyl ether.
At time of analytical testing a content of >100 ppm stabiliser was determined.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
40758555234808267
assay

≥99.0% (GC)

assay

-

assay

-

assay

-

solubility

1000 g/L

solubility

-

solubility

H2O: soluble

solubility

-

storage temp.

15-25°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

density

1.05 g/cm3 at 20 °C

density

1.16 g/mL at 25 °C (lit.)

density

1.214 g/mL at 25 °C (lit.)

density

-

form

liquid

form

liquid

form

-

form

-

bp

141 °C/1013 hPa

bp

-

bp

103 °C/19 mmHg (lit.)

bp

125 °C/25 mmHg (lit.)


Still not finding the right product?

Explore all of our products under Acrylic acid


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

119.3 °F - closed cup

flash_point_c

48.5 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters