Skip to Content
Merck

Skip To

1.10866

WL nutrient agar

GranuCult® plus, according to ISO 10718, granular, selective for yeasts, pack of 500 g

Synonym(s):

Wallerstein Laboratories Nutrient Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.74
UNSPSC Code:
41106213
Manufacturer/tradename:
GranuCult® plus
Agency:
according to ISO 10718
Ph:
5.3-5.7 (25 °C, 77 g/L in H2O, after autoclaving)
Form:
granular
Suitability:
selective for yeasts
Application(s):
food and beverages
microbiology
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

according to ISO 10718

Quality Segment

form

granular

packaging

pack of 500 g

manufacturer/tradename

GranuCult® plus

storage condition

dry media (Keep Tightly closed)

technique(s)

microbiological culture: suitable

color

blue-green

pH

5.3-5.7 (25 °C, 77 g/L in H2O, after autoclaving)

solubility

77 g/L

bulk density

730 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

selective for yeasts

General description

GranuCult® plus WL (Wallerstein Laboratories) Nutrient Agar has a pH of 5.5, which is optimal for the enumeration of brewers′ yeast. If bakers′ or distillers′ yeast is to be examined, the pH should be adjusted to 6.5 (better yields). When cultivating microorganisms from an alcoholic mash, tomato juice should be added to the medium. WL differential agar contains cycloheximide to suppress yeasts and any molds that may be present; this medium allows reliable counting of all bacteria that may be encountered in the tests performed in brewery laboratories.

Application

GranuCult® plus WL nutrient agar is suitable for the enumeration and cultivation of yeasts and bacteria in the microbiological control conducted in the brewing and other fermentation industries.

Features and Benefits

  • GranuCult® offers superior granulated culture media.
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment.
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions.
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Analysis Note

Appearance (clearness): clear
Appearance (colour): blue-green
pH-value (25 °C): 5.3 - 5.7
Growth (Candida albicans ATCC 10231 (WDCM 00054)): good to very good
Growth (Saccharomyces cerevisiae ATCC 9763 (WDCM 00058)): good to very good
Growth (Lactobacillus acidophilus ATCC 4356 (WDCM 00098)): fair to very good
Growth (Leuconostoc mesenteroides ATCC 9135 (WDCM 00108)): good to very good
Growth (Enterococcus faecalis ATCC 11700): good to very good
Growth (Escherichia coli ATCC 11775 (WDCM 00090)): good to very good
Growth (Proteus mirabilis ATCC 29906 (WDCM 00023)): good to very good
Incubation: 4 days; 28 °C; aerobic.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect®culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.114711.0061004163
suitability

yeasts

suitability

heterotrophic bacteria

suitability

bacteria, yeasts

suitability

nonselective for bacteria (General Media)

form

medium granules (dehydrated (DCM))

form

granular

form

-

form

powder

application(s)

food and beverages, microbiology

application(s)

food and beverages
microbiology

application(s)

food and beverages
microbiology

application(s)

agriculture
bioburden testing
environmental
food and beverages
life science and biopharma
water monitoring, microbiology

manufacturer/tradename

GranuCult® plus

manufacturer/tradename

NutriSelect® Prime

manufacturer/tradename

-

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

pH

5.3-5.7 (25 °C, 77 g/L in H2O, after autoclaving)

pH

7.1-7.5 (37 °C, 40 g/L in H2O, after autoclaving)

pH

-

pH

-


Still not finding the right product?

Explore all of our products under WL nutrient agar


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
110866050004022536128283

Questions

Reviews

No rating value

Active Filters