Skip to Content
Merck

Skip To

1.08643

Triton X-100

EMPROVE® EXPERT Ph Eur

Synonym(s):

Octoxynol, (p-tert-Octylphenoxy)polyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
UNSPSC Code:
12352104
Beilstein/REAXYS Number:
2315025
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

Ph. Eur.

Quality Level

vapor pressure

<0.01 hPa ( 20 °C)

product line

EMPROVE® EXPERT

form

liquid

potency

1900-5000 mg/kg LD50, oral (Rat), >3000 mg/kg LD50, skin (Rabbit)

mol wt

micellar avg mol wt 80,000, average mol wt 625

aggregation number

100-155

pH

5.0-8.0 (20 °C, 10 g/L in H2O)

CMC

0.2-0.9 mM (20-25°C)

bp

>200 °C/1013 hPa

transition temp

flash point 251 °C, cloud point 65 °C, pour point ~7 °C

density

1.07 g/cm3 at 20 °C

HLB

13.5

storage temp.

15-25°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

General description

Finding the right chemical that matches your pharma and biopharma manufacturing needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for pharma and biopharma manufacturing withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Other Notes

Due to its low microbial and endotoxin limits, Triton X-100 Emprove® Expert is suitable for biopharma manufacturing needs.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
X100T9284X100PC
form

liquid

form

liquid

form

liquid

form

liquid

product line

EMPROVE® EXPERT

product line

-

product line

BioXtra

product line

-

agency

Ph. Eur.

agency

-

agency

-

agency

-

mol wt

micellar avg mol wt 80,000, average mol wt 625

mol wt

micellar avg mol wt 80,000, average mol wt 625

mol wt

micellar avg mol wt 80,000, average mol wt 625

mol wt

micellar avg mol wt 80,000, average mol wt 625

density

1.07 g/cm3 at 20 °C

density

-

density

1.065 g/cm3 at 20 °C (68 °F)

density

-

bp

>200 °C/1013 hPa

bp

-

bp

-

bp

-


Still not finding the right product?

Explore all of our products under Triton X-100


signalword

Danger

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

483.8 °F - closed cup

flash_point_c

251 °C - closed cup

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - ED ENV 1 - Eye Dam. 1 - Skin Irrit. 2



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Btissame El Hassouni et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-11)
Inhibition of ribosome biogenesis has recently emerged as a promising strategy for the treatment of metastatic tumors. The RNA polymerase I inhibitor CX-5461 has shown efficacy in a panel of cancer types and is currently being tested in clinical trials.



Global Trade Item Number

SKUGTIN
108643902704027269222536
108643250004022536871622
108643602504027269078621
108643619004022536961743
108643916804027269222543
108643100004022536878416

Questions

Reviews

No rating value

Active Filters