Skip to Content
Merck

Skip To

1.06002

Methanol

≥99.9% (GC), suitable for UV/Vis spectroscopy, Uvasol®

Synonym(s):

Methyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
NACRES:
NA.03
EC Index Number:
200-659-6
UNSPSC Code:
12191502
MDL number:
Beilstein/REAXYS Number:
1098229
eCl@ss:
39020101
Assay:
≥99.9% (GC)
Technique(s):
UV/Vis spectroscopy: suitable
Bp:
64.7 °C (lit.)
Vapor pressure:
128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Methanol, for spectroscopy Uvasol®

vapor density

1.11 (vs air)

Quality Segment

vapor pressure

128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)

assay

≥99.9% (GC)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat), 17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity, ≤0.0002 meq/g Alkalinity, ≤0.01% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

205 nm, ≥10%, 210 nm, ≥30%, 220 nm, ≥60%, 230 nm, ≥80%, 240 nm, ≥90%, 250 nm, ≥95%, 260 nm, ≥98%

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

UV absorption

λ: 205 nm Amax: ≤1.00, λ: 210 nm Amax: ≤0.52, λ: 220 nm Amax: ≤0.22, λ: 230 nm Amax: ≤0.10, λ: 240 nm Amax: ≤0.05, λ: 250 nm Amax: ≤0.02, λ: 260 nm Amax: ≤0.01

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Application



  • A simple and green offline-online capillary electrophoresis stacking strategy for the simultaneous determination of hydrophobic compounds in complicated samples using sodium dodecyl sulfate as the solubilizer and pseudophase.: This study presents a green offline-online capillary electrophoresis strategy for hydrophobic compounds, employing methanol as part of the solubilizing system. (Qin et al., 2024).


Analysis Note

Evaporation Number: 1.9
Purity (GC): ≥ 99.9 %
Evaporation residue: ≤ 0.0002 %
Water: ≤ 0.01 %
Color: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 1.0 ppb
Transmission (at 205nm): ≥ 10 %
Transmission (at 210 nm): ≥ 30 %
Transmission (at 220 nm): ≥ 60 %
Transmission (at 230 nm): ≥ 80 %
Transmission (at 240 nm): ≥ 90 %
Transmission (at 250 nm): ≥ 95 %
Transmission (from 260 nm): ≥ 98 %
Absorbance (at 205nm): ≤ 1.00
Absorbance (at 210 nm): ≤ 0.52
Absorbance (at 220 nm): ≤ 0.22
Absorbance (at 230 nm): ≤ 0.10
Absorbance (at 240 nm): ≤ 0.05
Absorbance (at 250 nm): ≤ 0.02
Absorbance (from 260 nm): ≤ 0.01
Meet the requirements of "Specific use spectroscopy" according ACS and Reag. Ph Eur

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.133511.060181.06011
assay

≥99.9% (GC)

assay

≥99.8% (GC)

assay

≥99.8% (GC)

assay

≥99.8% (GC)

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

technique(s)

gas chromatography (GC): suitable

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

density

0.791 g/mL at 25 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

vapor density

1.11 (vs air)

vapor density

1.11 (vs air)

vapor density

1.11 (vs air)

vapor density

1.11 (vs air)


Still not finding the right product?

Explore all of our products under Methanol


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup

target_organs

Eyes,Central nervous system



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
106002050004022536070940
106002250004022536070964

Questions

Reviews

No rating value

Active Filters