Skip to Content
Merck

Skip To

1.04367

Hexane

≥99% (GC), ACS reagent, EMSURE®

Synonym(s):

Hexane, n-Hexane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
UNSPSC Code:
12191502
EC Index Number:
203-777-6
NACRES:
NB.21
Beilstein/REAXYS Number:
1730733
MDL number:
Assay:
≥99% (GC)
Grade:
ACS reagent
Bp:
69 °C (lit.)
Vapor pressure:
256 mmHg ( 37.7 °C), 5.2 psi ( 37.7 °C), ~132 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

n-Hexane, for analysis EMSURE® ACS

grade

ACS reagent

Quality Level

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C), 5.2 psi ( 37.7 °C), ~132 mmHg ( 20 °C)

product line

EMSURE®

assay

≥99% (GC)

form

liquid

autoignition temp.

453 °F

potency

25000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7.7 %

dilution

(for analytical testing)

impurities

≤0.0002 meq/g Acidity, ≤0.0002 meq/g Alkalinity, ≤0.005% Sulfur compounds (as S), ≤0.005% Water, ≤0.01% Aromatics (as benzene)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.375 (lit.)

kinematic viscosity

0.50 cSt(20 °C)

bp

69 °C (lit.)

mp

−95 °C (lit.)

transition temp

flash point -22 °C

density

0.659 g/mL at 25 °C (lit.)

SMILES string

CCCCCC

cation traces

Al: ≤0.00005%, B: ≤0.000002%, Ba: ≤0.00001%, Ca: ≤0.00005%, Cd: ≤0.000005%, Co: ≤0.000002%, Cr: ≤0.000002%, Cu: ≤0.000002%, Fe: ≤0.00001%, Mg: ≤0.00001%, Mn: ≤0.000002%, Ni: ≤0.000002%, Pb: ≤0.00001%, Sn: ≤0.00001%, Zn: ≤0.00001%

storage temp.

2-30°C

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application

n-Hexane is used:
  • as a washing solvent for microcapsules.
  • in reaction medium during polymerizations as a diluent for quick-drying paints, printing inks and adhesives.

Other Notes

Discover EMSURE - to be more than sure!
Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.070231.043711.04391
Quality Level

300

Quality Level

200

Quality Level

100

Quality Level

100

grade

ACS reagent

grade

ACS reagent

grade

-

grade

-

assay

≥99% (GC)

assay

≥98.5% (Σ hexane isomers + methylcyclopentane )

assay

≥98% (GC)

assay

≥98% (GC)

form

liquid

form

liquid

form

liquid

form

liquid

cation traces

Al: ≤0.00005%, Ba: ≤0.00001%, Cd: ≤0.000005%, Cr: ≤0.000002%, Fe: ≤0.00001%, Mn: ≤0.000002%, Pb: ≤0.00001%, Zn: ≤0.00001%, B: ≤0.000002%, Co: ≤0.000002%, Mg: ≤0.00001%, Sn: ≤0.00001%, Ca: ≤0.00005%, Ni: ≤0.000002%, Cu: ≤0.000002%

cation traces

-

cation traces

-

cation traces

-

impurities

≤0.0002 meq/g Acidity, ≤0.005% Sulfur compounds (as S), ≤0.01% Aromatics (as benzene), ≤0.0002 meq/g Alkalinity, ≤0.005% Water

impurities

≤0.0003 meq/g Water-soluble titrable acid, ≤0.005% Sulfur compounds (as S), ≤0.02% Water

impurities

-

impurities

≤0.0002 meq/g Acidity, ≤0.0002 meq/g Alkalinity, ≤0.01% Water


Still not finding the right product?

Explore all of our products under n-Hexane


signalword

Danger

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

flash_point_f

-7.6 °F - closed cup

flash_point_c

-22 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Release of thyme oil from polylactide microcapsules
Isabel M M et al.
Industrial & Engineering Chemistry Research, 50, 13752-13761 (2011)



Global Trade Item Number

SKUGTIN
104367250004022536050539
104367000004022536050508
104367602504022536050560
104367251104027269060220
104367100004022536050515
104367619004022536719979

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters