Skip to Content
Merck

Skip To

1.02735

L-Cysteine hydrochloride monohydrate

EMPROVE® EXPERT, Ph. Eur., JP, USP

Synonym(s):

Cys, L-Cysteinium Chloride Monohydrate, L-Cysteine Hydrochloride, Monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
EC Index Number:
200-157-7
UNSPSC Code:
12352209
Beilstein/REAXYS Number:
5158059
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

JP, Ph. Eur., USP

vapor pressure

<0.1 hPa ( 20 °C)

product line

EMPROVE® EXPERT

assay

98.5-101.0% dry basis (alkalimetric)

form

crystalline powder

pH

0.8-1.2 (20 °C, 100 g/L in H2O)

mp

168-170 °C

solubility

650 g/L

density

1.54 g/cm3

bulk density

780 kg/m3

anion traces

sulfate (SO42-): ≤300 ppm

cation traces

Fe: ≤20 ppm, heavy metals (as Pb): ≤10 ppm

storage temp.

2-30°C

SMILES string

O.Cl.N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2/t2-;;/m0../s1

InChI key

QIJRTFXNRTXDIP-JIZZDEOASA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Biochem/physiol Actions

NMDA glutamatergic receptor antagonist.

Analysis Note

Assay (alkalimetric, calculated on dried substance): 98.5 - 101.0 %
Identity (IR-spectrum): passes test
Identity (Chloride): passes test
Appearance: white to almost white crystalline powder or colorless crystals
Appearance of solution (25 g/l, water): clear and not more intense in color than reference solution BY₆
Spec. rotation (α 20/D, 80 g/l, hydrochloric acid 250 g/l, calc. on dried substance): +5.5 to +7.0
Spec. rotation (α 25/D, 80 g/l, hydrochloric acid 6 mol/l, calc. on dried substance): +5.7 to +6.8
Sulfate (SO₄): ≤ 300 ppm
Heavy metals (as Pb): ≤ 10 ppm
Fe (Iron): ≤ 20 ppm
Related Compounds, total (TLC): ≤ 0.5 %
Ninhydrin-positive substances (LC)(impurity A (570 nm)): ≤ 0.5 %
Ninhydrin-positive substances (LC)(any ninhydrin-positive impurity): ≤ 0.2 %
Ninhydrin-positive substances (LC) (ammonium (570 nm)): ≤ 0.02 %
Ninhydrin-positive substances (LC)(total impurities): ≤ 1.0 %
Residual solvents (ICH Q3C): excluded by manufacturing proces
Sulfated ash (600 °C): ≤ 0.1 %
Loss on drying (Vacuum < 0.7 kPa, P₂O₅, 24 h): 8.0 - 12.0 %
Bacterial endotoxins: ≤ 6.0 I.U./g
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph. Eur., USP.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1102C6852C7880
agency

JP, USP, Ph. Eur.

agency

traceable to Ph. Eur. C3290000, traceable to USP 1161509

agency

meets EP testing specifications, meets USP testing specifications

agency

-

form

crystalline powder

form

-

form

powder or crystals

form

powder

assay

98.5-101.0% dry basis (alkalimetric)

assay

-

assay

98.5-101.0% dry basis

assay

≥98% (TLC)

product line

EMPROVE® EXPERT

product line

-

product line

-

product line

-

solubility

650 g/L

solubility

-

solubility

water: 1 g/10 mL, clear, colorless

solubility

-

density

1.54 g/cm3

density

-

density

-

density

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
102735100004022536942629
102735901004022536942636
102735010004022536942612
102735902504054839392207
102735000104054839392191

Questions

Reviews

No rating value

Active Filters