Skip to Content
Merck

Skip To

1.02402

Iron(II) ethylenediammonium sulfate

Volumetric standard, secondary reference material for redox titration, traceable to NIST Standard Reference Material (SRM) Certipur® Reag. Ph Eur

Synonym(s):

Iron(II) ethylenediammonium sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
EC Index Number:
264-357-6
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

agency

ISO 17025, reag. Ph. Eur.

form

solid

technique(s)

titration: suitable

pH

4.0 (25 °C, 100 g/L in H2O)

solubility

170 g/L

bulk density

600 kg/m3

storage temp.

15-25°C

SMILES string

O=S(=O)(O)O.O.[FeH2].NCCN

InChI

1S/C2H8N2.Fe.H2O4S/c3-1-2-4;;1-5(2,3)4/h1-4H2;;(H2,1,2,3,4)/q;+3;

InChI key

WAYQJNDEVYLLFJ-UHFFFAOYSA-N

General description

Iron(II) ethylenediammonium sulfate volumetric standard is manufactured under stringent conditions and measured with the highest possible precision. Its mass fraction is determined by potentiometric titration, and it is qualified in a DIN EN ISO/IEC 17025 accredited laboratory.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It finds its use as a certified reference material (CRM) for the standardization of volumetric solutions in redox titrations, according to USP and Ph. Eur.

Features and Benefits

  • Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • Certified reference material according to ISO 17034
  • High-purity material traceable to NIST SRMs
  • Qualified in a DIN EN ISO/IEC 17025 accredited laboratory
  • In accordance with the chapter reagents of USP and Ph. Eur
  • High-quality offering accurate titer determinations

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2035051.03963414913
technique(s)

titration: suitable

technique(s)

-

technique(s)

-

technique(s)

-

Quality Level

300

Quality Level

200

Quality Level

500

Quality Level

100

bulk density

600 kg/m3

bulk density

-

bulk density

-

bulk density

-

storage temp.

15-25°C

storage temp.

-

storage temp.

15-25°C

storage temp.

-

form

solid

form

crystalline

form

crystalline, solid

form

solid

agency

ISO 17025, reag. Ph. Eur.

agency

-

agency

BP, Ph. Eur., USP

agency

-


Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
102402008304054839568695
102402008004022536555782

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters