Skip to Content
Merck

Skip To

1.01352

May-Grünwald′s eosin methylene blue

for microscopy

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.21
UNSPSC Code:
41116130
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

solid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

bulk density

530 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

General description

The May-Grünwald′s eosin methylene blue staining dye for microscopy, is used to make a stain that is frequently used for diagnostic purposes in the area of hematology. When used in hematological applications, May-Grünwald′s stain is frequently used in combination with other staining solutions, e. g. with Giemsa′s solution for Pappenheim (MGG) overview staining. This staining solution generally stains the nuclei red to violet, based on the molecular interaction between the Eosin Y dye and an Azure B-DNA complex. Both dyes assemble to an Eosin Y - Azure B-DNA complex and the intensity of the resulting stain depends on the content of Azure B and the ratio of Azure B : Eosin Y. Furthermore, the resulting stain can vary depending on the influence of fixation, staining times, pH-value of the solutions or buffer substances. As a CE-certified, IVD-registered product, it can be used in diagnostics and laboratory accreditations without constraints. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.014242054351.09203
technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

form

solid

form

liquid

form

powder

form

solid

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

-

IVD

for in vitro diagnostic use

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

room temp

storage temp.

2-30°C

Quality Level

400

Quality Level

400

Quality Level

200

Quality Level

400


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
101352002504022536019710
101352100004022536019734
101352010004022536019727

Questions

Reviews

No rating value

Active Filters